8.22084
L-(+)-Lithium lactate
≥98%
Synonym(s):
L-(+)-Lithium lactate, Lactic acid lithium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3CH(OH)COOLi
CAS Number:
Molecular Weight:
96.01
MDL number:
UNSPSC Code:
12352107
EC Index Number:
248-692-5
NACRES:
NA.22
Recommended Products
General description
for synthesis
Application
- Determination of lactate dehydrogenase in human erythrocytes by capillary electrophoresis with electrochemical detection.: This study explores the use of L-(+)-Lithium lactate in the determination of lactate dehydrogenase activity in human erythrocytes using capillary electrophoresis with electrochemical detection. The research highlights the compound′s effectiveness in providing precise measurements, crucial for biochemical and clinical applications (Wang et al., 2003).
Analysis Note
Assay (HClO₄): ≥ 98.0 %
Spec. rotation [α²0/D (c=4 in water): -15.0 - -13.0 °
Identity (IR): passes test
Spec. rotation [α²0/D (c=4 in water): -15.0 - -13.0 °
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service