Skip to Content
Merck
All Photos(3)

Documents

440469

Sigma-Aldrich

Lithium lactate

95%

Synonym(s):

Lactic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOLi
CAS Number:
Molecular Weight:
96.01
Beilstein:
3568332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

>300 °C (lit.)

solubility

water: soluble(lit.)

SMILES string

[Li+].CC(O)C([O-])=O

InChI

1S/C3H6O3.Li/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1

InChI key

GKQWYZBANWAFMQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium lactate may be employed in the following studies:
  • As an electron spin resonance (ESR) dosimeter material.
  • As lactic acid standard in the determination of lactic acid.
  • As lactic acid standard in the determination of lactic acid with muscle lactic dehydrogenase by an enzymatic method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The determination of lactic acid.
Friedemann TE and Graeser JB.
The Journal of Biological Chemistry, 100(1), 291-308 (1933)
Optimal conditions for the enzymatic determination of L-lactic acid.
G F OLSON
Clinical chemistry, 8, 1-10 (1962-02-01)
Lithium lactate as an ESR dosimeter.
Hassan GM, et al.
Applied Radiation and Isotopes, 49(7), 823-828 (1998)
Bettina Ziegelmann et al.
Scientific reports, 8(1), 683-683 (2018-01-14)
Honey bees are increasingly important in the pollination of crops and wild plants. Recent reports of the weakening and periodical high losses of managed honey bee colonies have alarmed beekeeper, farmers and scientists. Infestations with the ectoparasitic mite Varroa destructor
E Brindha et al.
Toxicology mechanisms and methods, 25(2), 150-154 (2015-01-07)
This study was aimed to evaluate the preventive role of phytic acid on lysosomal enzymes in isoproterenol (ISO)-induced myocardial infarction (MI) in male Wistar rats. Rats subcutaneously injected with ISO (85 mg/kg) at an interval of 24 h for two days showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service