251380
L-(+)-Tartaric acid
ACS reagent, ≥99.5%
Synonym(s):
(2R,3R)-(+)-Tartaric acid, L-Threaric acid
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
suitable for EPA 200.7
suitable for EPA 200.8
vapor density
5.18 (vs air)
Assay
≥99.5%
form
crystalline powder
crystals
optical activity
[α]20/D +12.4°, c = 20 in H2O
optical purity
ee: 99% (GLC)
autoignition temp.
797 °F
impurities
≤0.002% S compounds
≤0.005% insolubles
ign. residue
≤0.02%
mp
170-172 °C (lit.)
anion traces
chloride (Cl-): ≤0.001%
oxalate (C2O42-): passes test
phosphate (PO43-): ≤0.001%
cation traces
Fe: ≤5 ppm
heavy metals: ≤5 ppm (by ICP-OES)
functional group
carboxylic acid
hydroxyl
SMILES string
O[C@H]([C@@H](O)C(O)=O)C(O)=O
InChI
1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1
InChI key
FEWJPZIEWOKRBE-JCYAYHJZSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
302.0 °F - closed cup
Flash Point(C)
150 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service