Skip to Content
Merck
All Photos(7)

Documents

8.18760

Supelco

Ethyl alcohol, pure

greener alternative

≥99.5% (GC), for preparative purposes, synthesis, cleaning, extraction and analytical purposes, EMPLURA®

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-578-6
NACRES:
NB.21

product name

Ethanol, absolute EMPLURA®

grade

for analytical purposes
for cleaning
for extraction
for preparative purposes
for synthesis

Quality Level

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)
59 hPa ( 20 °C)

product line

EMPLURA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

683 °F

potency

6200 mg/kg LD50, oral (Rat)

expl. lim.

19 %, 60 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.2% Water

evapn. residue

≤0.0025%

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−114 °C (lit.)

transition temp

flash point 12 °C

density

0.789 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical

format

neat

greener alternative category

storage temp.

2-30°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanol (EMPLURA® grade product) is ideal for numerous basic applications, such as preparative lab work, cleaning, or standard production processes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biorenewable and alternative to petrochemical solvents. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here  for more information.

Application


  • Effects of ethanol or ethylene glycol exposure on PPAR gamma and aromatase expression in adipose tissue.: This study explores the biochemical effects of ethanol exposure on protein expressions impacting metabolic pathways in adipose tissues, highlighting its analytical application in understanding chemical interactions within biological systems (Vogel et al., 2024).

  • Advancing ethanol content determination in hydrogels: non-destructive and operational methods for health and criminal inspections.: Focuses on non-destructive techniques for determining ethanol content in hydrogels, critical for applications in healthcare monitoring and forensic analysis, offering insights into ethanols role in public health and safety (de O Morais et al., 2024).

  • The gastroprotective effect of Yucca filamentosa standardized crude leaves extract versus its nano-cubosomal formulation in ethanol-induced gastric injury.: Investigates the protective effects of plant extracts against ethanol-induced gastric damage, utilizing ethanol as a tool for studying therapeutic responses and formulation efficacy in clinical biochemistry settings (Hammad et al., 2024).

  • Atomic Layer Deposition of TiO(2) on Si Window Enables In Situ ATR-SEIRAS Measurements in Strong Alkaline Electrolytes.: Describes the enhancement of spectroscopic measurements through the atomic layer deposition of TiO2, facilitated by ethanol processes, improving analytical methodologies in chemically aggressive environments (Cai et al., 2024).

Features and Benefits

This bioethanol is a suitable choice for a sustainable future because of its high quality, affordability, and ready availability.     
  • It is produced from grain or sugarcane, a renewable source
  • Less toxic than synthetic ethanol (no toxic by-products)     
  • The production method is safer for the environment

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.789 - 0.790
Evaporation residue: ≤ 0.0025 %
Water: ≤ 0.20 %

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service