Skip to Content
Merck
All Photos(1)

Documents

1.08543

Sigma-Aldrich

Ethyl alcohol, pure

≥99.8% (GC), for molecular biology

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-578-6
NACRES:
NA.22

product name

Ethanol, for molecular biology

vapor density

1.59 (vs air)

Quality Level

vapor pressure

44.6 mmHg ( 20 °C)
59 hPa ( 20 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

683 °F

potency

6200 mg/kg LD50, oral (Rat)

expl. lim.

19 %, 60 °F

impurities

DNAses (nicking-activity), none detected
RNAses, none detected
≤0.0001% Heavy metals (as Pb)
≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity

color

colorless

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−114 °C (lit.)

transition temp

flash point 12 °C

density

0.789 g/mL at 25 °C (lit.)

format

neat

storage temp.

15-25°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for molecular biology

Application

Ethanol for molecular biology. CAS No. 64-17-5, EC Number 200-578-6.

Analysis Note

Appearance (colour)colourlessAppearance (description)clearPurity (GC)≥ 99.8 %Acidity≤ 0.0002 meq/gAlkalinity≤ 0.0002 meq/gHeavy metals (as Pb)≤ 0.0001 %DNases (nicking-activity)not detectableRNasesnot detectable

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service