Skip to Content
Merck
Ethanol tested according to Ph. Eur.

Ethyl alcohol, Pure

Ethanol tested according to Ph. Eur.
Synonym(s):
Ethanol, absolute alcohol, non-denatured ethanol, Ethyl alcohol
Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing