71300-M
Overnight Express Autoinduction System 1 - Novagen
The Overnight Express Autoinduction System allows the induction of protein expression without monitoring cell density and without conventional induction with IPTG.
Synonym(s):
autoinduction media, autoinduction medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106212
NACRES:
NA.54
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Novagen®
storage condition
OK to freeze
technique(s)
protein expression: suitable
protein purification: suitable
storage temp.
10-30°C
Related Categories
General description
The Overnight Express™ Autoinduction System allows the induction of protein expression without monitoring cell density and without conventional induction with isopropyl β-D-1-thiogalactopyranoside (IPTG). This system is extremely convenient for routine expression of proteins in multiple cultures and is ideal for high-throughput parallel analysis of protein expression, solubility, and purification from multiple expression clones. The Overnight Express Autoinduction System together with the Novagen RoboPop Solubility Screening Kit and RoboPop Purification Kits allows rapid optimization of the host-vector combinations, expression conditions, and purification parameters for high-throughput production of proteins for structural or functional analysis.
Application
Overnight Express™ Autoinduction System 1 - Novagen has been used for the purification of 15N-human prion proteins (HuPrPs). It is suitable for most routine recombinant protein expression applications.
Features and Benefits
- No need to monitor cell growth or add IPTG
- High cell densities and protein expression levels
- Increased soluble protein yield
- Induction of numerous expression clones simultaneously
- Usable with any conventional glucose-free bacterial growth medium
- Ideal for pET Expression System or other IPTG-inducible bacterial systems
- Compatible with cultures grown in flasks, culture tubes, and deep-well plates
- Minimal sample handling
Components
The Overnight Express™ Autoinduction System 1 kit contains three concentrated sterile solutions:
- OnEx Solution 1 A blend of carbon sources optimized for tightly regulated uninduced growth to high cell density, followed by high-level induction
- OnEx Solution 2 A concentrated buffer and nitrogen-source bend that mediates metabolic acid production and provides additional nitrogen for increased protein synthesis
- OnEx Solution 3 Provides magnesium for maximal cell density
Warning
Toxicity: Standard Handling (A)
Other Notes
Grabski, A., et al. 2003. inNovations 17, 3.
Legal Information
Express is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Andrew G Reidenbach et al.
The Journal of biological chemistry, 295(39), 13516-13531 (2020-07-30)
Prion disease is a rapidly progressive neurodegenerative disorder caused by misfolding and aggregation of the prion protein (PrP), and there are currently no therapeutic options. PrP ligands could theoretically antagonize prion formation by protecting the native protein from misfolding or
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service