Skip to Content
Merck

Skip To

E1014

Benzonase® Nuclease

≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution

Synonym(s):

Endonuclease from Serratia marcescens

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Assay:
≥90% (SDS-PAGE)
Biological source:
Serratia marcescens
Recombinant:
expressed in E. coli
Concentration:
≥250 units/μL
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Serratia marcescens

Quality Segment

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

30 kDa

concentration

≥250 units/μL

input

general use sample(s)

application(s)

research use

foreign activity

protease, essentially free

shipped in

wet ice

storage temp.

−20°C

General description

Benzonase® nuclease is a highly efficient and genetically engineered endonuclease that originates from Serratia marcescens. This dimeric protein with two essential disulfide bonds is capable of attacking and degrading all forms of DNA and RNA (single-stranded, double-stranded, linear, and circular) under a wide range of operating conditions. Benzonase® nuclease is capable of removing nucleic acids and enhancing the purity and quality of protein samples.

Application

Used for the removal of nucleic acid from protein samples.
Benzonase® Nuclease has been used: as a component in ice-cold lysis buffer C to digest DNA and  RNA to facilitate the complete release of all nuclear proteins in the immunoprecipitation step to release protein complexes from the nucleoplasm and chromatinas a supplement in RIPA to fractionate SHSY5Y cells for immunoprecipitation to remove residual nucleic acids from the aortic roots in decellularization method

Biochem/physiol Actions

Benzonase® Nuclease can completely digest nucleic acids into 5′-monophosphate terminated oligonucleotides of 3 to 5 bases in length, making it the ideal tool for removing nucleic acids from recombinant proteins and for applications that require complete digestion of nucleic acids. In addition to reducing viscosity in protein extracts and preventing cell clumping, pretreatment of protein samples with Benzonase® nuclease can significantly improve their resolution on 2D gel electrophoresis by eliminating any bound nucleic acids. This versatile enzyme can digest both native or heat-denatured DNA and RNA, with its optimum pH for enzyme activity found to be 8.0-9.2. Benzonase® nuclease is effective at removing host DNA from microbiome samples. In many cases, microbiome samples (such as saliva or skin) will have a high percentage of host DNA that interferes with downstream results. Our experts show that the reduction of host DNA lowers the cost of sequencing while increasing and improving the data. Experimental data is shown in the technical article - Benzonase® Nuclease for Microbiome Workflows
Digests native or heat-denatured DNA and RNA.

Features and Benefits

  • Host DNA depletion in microbiome samples.


  • Effective nucleic acid digestion in a variety of workflows.


  • Viscosity reduction during protein extraction.

Physical form

Solution in 50% glycerol containing 20 mM Tris HCl, pH 8.0, 2 mM MgCl2, and 20 mM NaCl.

Other Notes

One unit will digest sonicated salmon sperm DNA to acid-soluble oligonucleotides equivalent to a ΔA260 of 1.0 in 30 min at pH 8.0 at 37 °C (reaction volume 2.625 ml).

Legal Information

Benzonase® Nuclease is supplied by Merck KGaA, Darmstadt, Germany and/or its affiliates.
Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7066471206SRE0015
description

≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution

description

Effective viscosity reduction and removal of nucleic acids from protein solutions

description

Effective Viscosity Reduction and Removal of Nucleic Acids from protein solutions

description

recombinant, expressed in Pichia pastoris

assay

≥90% (SDS-PAGE)

assay

>99% (SDS-PAGE)

assay

>99% (SDS-PAGE)

assay

-

biological source

Serratia marcescens

biological source

Serratia marcescens

biological source

Serratia marcescens

biological source

fermentation/recombinant

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

application(s)

research use

application(s)

research use

application(s)

research use

application(s)

cell analysis

concentration

≥250 units/μL

concentration

25-29 units/μL

concentration

≥250 units/μL

concentration

-

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in Pichia pastoris


Still not finding the right product?

Explore all of our products under Benzonase® Nuclease


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

ppe

Eyeshields, Gloves

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–9 of 9 Questions  
  1. Does this product have a tag (e.g His)

    1 answer
    1. This item does not contain a His-tag.

      Helpful?

  2. hi, I received E1014-5KU product. I couldn't find what solution should I dilute to get a concentration of 25U/ml. Also, the volume of the solution.

    1 answer
    1. The actual volume of the solution is determined by the lot-specific activity, which can be found in the Certificate of Analysis. For this product, there are 5,000 units available. If the lot has a specific activity of 335 U/?L, the volume will be approximately 14.9 ?L for 5,000 units. It is recommended to dilute the product in 20 mM Tris-HCl (pH 8.0), 2 mM MgCl?, and 20 mM NaCl.

      Helpful?

  3. Do you sell high salt lysis buffers that are compatible with benzonase? Do you have any written protocols for use of benzonase with high salt buffers?

    1 answer
    1. We do not have a branded "high-salt lysis buffer" off the shelf that is compatible with benzonase. However, we offer a specialized Benzonase® Salt Tolerant Endonuclease which is active at higher salt concentration, up to1M NaCl .
      https://www.sigmaaldrich.com/US/en/product/mm/e5014

      Helpful?

  4. I am doing a protein purification and would like to know if magnesium chloride is required in the lysis buffer when using the Benzonase. Also, would the activity of benzonase be affected when added to a lysis buffer containing EDTA

    1 answer
    1. A concentration of 1-2 mM of magnesium is required for the activity of this product. An EDTA concentration of 1 mM partially inhibits the activity of this product.

      Helpful?

  5. Hi, what is the ratio of E1014 : DNA and E1014 : RNA for an efficient acid nucleic removal?

    1 answer
    1. For efficient nucleic acid removal using Benzonase® (E1014), the recommended ratio depends on the concentration of DNA or RNA in the sample. A typical starting point is 25 U/mL of Benzonase® for reducing nucleic acid content in lysates, which corresponds to approximately 1 unit of enzyme per 37 µg of DNA under optimal conditions. For lower concentrations of DNA or RNA, as little as 9 U/mL may achieve 99% removal, but higher concentrations, e.g., 90 U/mL, ensure faster and more complete degradation. Optimal activity requires 1–2 mM magnesium ions, a pH of 8.0–9.2, and incubation at 37°C. Adjustments may be needed for sample-specific factors such as buffer composition and nucleic acid load.

      Helpful?

  6. How many ul does it contain? What kind of Unit is KU?

    1 answer
    1. The unit activity of this product is lot-specific and reported in the product Certificate of Analysis. The minimum activity is 250 units per microliter. The KU value represents 1000 units. For example, a 20 KU package size represents 20,000 units. Please see the link below to review a sample or lot-specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/e1014#product-documentation

      Helpful?

  7. Hi, for protein purification from E. coli cells, how much is the working concentration range for Benzonase nuclease (≥250 units/μL)? Thank you 

    1 answer
    1. The recommended starting concentration for Benzonase nuclease is 25 units per milliliter of cell lysate.

      Helpful?

  8. Hello, I plan to do proteomic for protein. When I add RIPA buffer to isolate the protein, I need to remove the DNA and RNA inside. When should I add E1014? Together with RIPA or after it?

    1 answer
    1. Concentrations greater than 1 mM EDTA will inhibit Benzonase activity, and RIPA buffer recipes typically contain EDTA at higher concentrations than 1 mM. Therefore, Benzonase should be used after removing EDTA from the lysed sample or consider using a different lysis solution that does not include EDTA in the formulation.

      Helpful?

  9. What is the storage temperature range for E1014? There is only a specified storage temperature, and not a range.

    1 answer
    1. This product is stored at freezer temperature, which is typically -20°C. An excepted range is -15 - -20°C.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review