Skip to Content
Merck
All Photos(2)

Key Documents

S2671

Sigma-Aldrich

Suramin sodium salt

≥98% (TLC)

Synonym(s):

Suramine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H34N6Na6O23S6
CAS Number:
Molecular Weight:
1429.17
Beilstein:
3694087
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (TLC)

form

powder

impurities

<15% water (Karl Fischer)

color

white to light brown

solubility

H2O: >10 mg/mL

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].Cc1ccc(cc1NC(=O)c2cccc(NC(=O)Nc3cccc(c3)C(=O)Nc4cc(ccc4C)C(=O)Nc5ccc(c6cc(cc(c56)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O)c2)C(=O)Nc7ccc(c8cc(cc(c78)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C51H40N6O23S6.6Na/c1-25-9-11-29(49(60)54-37-13-15-41(83(69,70)71)35-21-33(81(63,64)65)23-43(45(35)37)85(75,76)77)19-39(25)56-47(58)27-5-3-7-31(17-27)52-51(62)53-32-8-4-6-28(18-32)48(59)57-40-20-30(12-10-26(40)2)50(61)55-38-14-16-42(84(72,73)74)36-22-34(82(66,67)68)24-44(46(36)38)86(78,79)80;;;;;;/h3-24H,1-2H3,(H,54,60)(H,55,61)(H,56,58)(H,57,59)(H2,52,53,62)(H,63,64,65)(H,66,67,68)(H,69,70,71)(H,72,73,74)(H,75,76,77)(H,78,79,80);;;;;;/q;6*+1/p-6

InChI key

VAPNKLKDKUDFHK-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Suramin is an antiparasitic drug, which blocks endothelial cell proliferation and increases tyrosine phosphorylation of many signalling proteins. It is associated with urticaria, paresthesias, vomiting and neurologic toxicity. Suramin is used to treat trypanosomiasis and onchocerciasis. It functions as an inhibitor of heparanase and also exhibits anti-AIDS (acquired immunodeficiency syndrome) property.

Application

Suramin sodium salt has been used:
  • as a potent blocker of rat P2Y2 purinergic receptor
  • to study its effects on short circuit current (Isc) across strial marginal cells
  • to show its effects on initial burst of glial activity
  • to assess its neuroprotective potential in rats
  • as a purinoceptor inhibitor, to explore the mechanism of action of extracellular adenosine triphosphate (eATP)

Biochem/physiol Actions

Suramin is a polysulfonated naphthylurea anticancer agent that inhibits tumor cell proliferation. It inhibits the activity of topoisomerase II by blocking the binding of the enzyme to DNA. It′s antiangiogenic activity may be related to its ability to bind to and inhibit the activity of several growth factors, including FGFa, FGFb, and PGDF. It uncouples G-proteins from receptors. It is an broad spectrum antagonist at P2X and P2Y purinergic receptors. Suramin has well documented antiprotozoal and anthelmintic activity.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drugs That Inhibit Signalling Pathways for Tumor Cell Growth and Proliferation
Medicinal Chemistry of Anticancer Drugs, 251-305 (2008)
Suramin reduces infarct volume in a model of focal brain ischemia in rats
Kharlamov A, et al.
Experimental Brain Research. Experimentelle Hirnforschung. Experimentation Cerebrale, 147(3), 353-359 (2002)
Inorganic polyphosphate regulates neuronal excitability through modulation of voltage-gated channels
Stotz SC, et al.
Molecular Brain, 7(1), 42-42 (2014)
Gabriela M Rozanski et al.
The European journal of neuroscience, 36(10), 3314-3321 (2012-08-01)
Somatic sensory neuron somata are located within the dorsal root ganglia (DRG) and are mostly ensheathed by individual satellite glial cell sheets. It has been noted, however, that a subpopulation of these DRG somata are intimately associated, separated only by
James M McCarthy et al.
PloS one, 8(1), e55282-e55282 (2013-02-06)
Prion diseases, also known as transmissible spongiform encephalopathies, are a group of fatal neurodegenerative diseases that include scrapie in sheep, bovine spongiform encephalopathy (BSE) in cattle and Creutzfeldt-Jakob disease (CJD) in humans. The 'protein only hypothesis' advocates that PrP(Sc), an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service