Skip to Content
Merck
All Photos(6)

Documents

1.08292

Supelco

2-Methyltetrahydrofuran

greener alternative

EMPLURA®

Synonym(s):

2-MeTHF, Tetrahydro-2-methylfuran, Tetrahydrosilvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102448
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-507-4
NACRES:
NA.05

vapor pressure

136 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

270 °C

potency

300-2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

1.2-5.7 % (v/v)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.406 (lit.)

bp

78-80 °C (lit.)

mp

-136 °C

transition temp

flash point -11 °C

solubility

150 g/L

density

0.86 g/mL at 25 °C (lit.)

greener alternative category

storage temp.

2-30°C

SMILES string

CC1CCCO1

InChI

1S/C5H10O/c1-5-3-2-4-6-5/h5H,2-4H2,1H3

InChI key

JWUJQDFVADABEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a bio-based and thus aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here for more information.

Features and Benefits

Greener alternative for THF, DCM, DMSO, and MTBE

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service