Skip to Content
Merck
All Photos(8)

Key Documents

1.08325

Supelco

Toluene

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Toluol, Methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-625-9
NACRES:
NA.21
grade:
ACS reagent
for analytical purposes
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

product line

EMSURE®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

dilution

(for analytical testing)

impurities

≤0.000002% As (Arsenic)
≤0.0002 meq/g Acidity
≤0.0006 meq/g Alkalinity
≤0.005% Benzene (GC)
≤0.03% Water
≤0.5 ppm Chloride (Cl)
≤1 ppm Sulfate (SO4)

evapn. residue

≤0.0005%

color

clearAPHA: ≤10

refractive index

n/D 1.496 (lit.)

kinematic viscosity

0.7 cSt(20 °C)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

transition temp

flash point 4 °C

solubility

0.52 g/L

density

0.865 g/mL at 25 °C (lit.)

cation traces

Ag: ≤0.000002%
Al: ≤0.00005%
Au: ≤0.00001%
B: ≤0.000002%
Ba: ≤0.00001%
Be: ≤0.000002%
Bi: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
In: ≤0.000002%
Li: ≤0.000002%
Mg: ≤0.00001%
Mn: ≤0.000002%
Mo: ≤0.000005%
Ni: ≤0.000002%
Pb: ≤0.00001%
Pt: ≤0.000002%
Sb: ≤0.000002%
Sn: ≤0.00001%
Ti: ≤0.000005%
Tl: ≤0.000005%
V: ≤0.000005%
Zn: ≤0.00001%
Zr: ≤0.000002%

SMILES string

Cc1ccccc1

storage temp.

2-30°C

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Toluene Detection in Biological Samples Using SPME-GC-MS.: This research presents a method for detecting toluene in biological samples using solid-phase microextraction combined with gas chromatography-mass spectrometry (SPME-GC-MS). The method provides high sensitivity and accuracy, making it suitable for biological monitoring of toluene exposure (Suh et al., 2015).

  • Adsorption Characteristics of Toluene on Metal-Organic Frameworks.: This paper explores the adsorption characteristics of toluene on various metal-organic frameworks (MOFs). The study identifies the most efficient MOFs for toluene adsorption, offering insights for industrial applications in air purification (Zhang et al., 2019).

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service