Skip to Content
Merck
All Photos(1)

Key Documents

1.01737

Supelco

Barium hydroxide octahydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Barium hydroxide octahydrate, Caustic baryta, Barium oxide hydrate octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OH)2 · 8 H2O
CAS Number:
Molecular Weight:
315.46
MDL number:
UNSPSC Code:
12352305
EC Index Number:
241-234-5
NACRES:
NA.21
Assay:
≥98.0% (acidimetric)
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

≥98.0% (acidimetric)

form

solid

potency

550 mg/kg LD50, oral (Rat)

impurities

≤0.005% Substances insoluble in dilute hydrochloric acid

pH

14 (20 °C in H2O, saturated aqueous solution)

mp

78 °C

density

2.18 g/cm3 at 20 °C

bulk density

900‑1100 kg/m3

anion traces

carbonate (as BaCO3): ≤2.0%
chloride (Cl-): ≤0.001%
sulfide (S2-): ≤0.0005%

cation traces

Ca: ≤0.002%
Fe: ≤0.0005%
K: ≤0.01%
Na: ≤0.01%
Pb: ≤0.001%
Sr: ≤0.5%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Ba+2].[O-H].[O-H].O.O.O.O.O.O.O.O

InChI

1S/Ba.10H2O/h;10*1H2/q+2;;;;;;;;;;/p-2

InChI key

ZUDYPQRUOYEARG-UHFFFAOYSA-L

Application


  • Determination of phosphorylated and O-glycosylated sites by chemical targeting (CTID) at ambient temperature.: This study explores the use of chemical targeting to identify phosphorylated and O-glycosylated sites, essential for understanding protein modifications. Barium hydroxide is utilized for its effectiveness in ambient temperature conditions, contributing to more efficient analytical methods (Hathaway, 2007).

  • Wet-chemical synthesis of crystalline BaTiO3 from stable chelated titanium complex: formation mechanism and dispersibility in organic solvents.: The research demonstrates the wet-chemical synthesis of BaTiO3, highlighting the formation mechanism and its dispersibility. Barium hydroxide plays a crucial role in stabilizing the chelated titanium complex, making it a valuable component in material synthesis and analytical chemistry (Pramanik et al., 2006).

  • Synthesis and analgesic effect of normorphine-3- and -6-glucuronides.: This paper details the synthesis of normorphine glucuronides, where barium hydroxide is used in the synthetic process. The study contributes to the understanding of analgesic properties and the chemical synthesis of pharmacologically relevant compounds (Oguri et al., 1989).



Analysis Note

Assay (acidimetric): ≥ 98.0 %
Substances insoluble in dilute hydrochloric acid: ≤ 0.005 %
Carbonate (as BaCO₃): ≤ 2.0 %
Chloride (Cl): ≤ 0.001 %
Sulphide (S): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.002 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.01 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.001 %
Sr (Strontium): ≤ 0.5 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service