Skip to Content
Merck
All Photos(1)

Documents

1.09992

Supelco

Titriplex® III solution

c(Na₂-EDTA 2 H₂O) = 0.1 mol/l, concentrate for preparing 1000 ml volumetric solution for titration, Titrisol®®

Synonym(s):

Complexometric titrant III

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.21

product name

Titriplex® III solution, for 1000 ml, c(Na₂-EDTA 2 H₂O) = 0.1 mol/l Titrisol®

Quality Level

product line

Titrisol®

form

liquid

concentration

0.1 M

technique(s)

titration: suitable

pH

7.4-7.6 (20 °C in H2O)

density

1.14 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • The use of Titriplex III for cross-matching for urgent blood transfusions and patients with blood coagulation disorders: Demonstrates the application of Titriplex III in critical healthcare settings, particularly in ensuring the compatibility of blood transfusions, crucial for emergency medical procedures (Fischer & Busch, 1960).

  • Clinical use of ethylenediaminetetracetic acid, sodium salt (Titriplex III) as an anticoagulant: Highlights the role of Titriplex III in enhancing the safety and effectiveness of clinical practices by preventing blood coagulation during medical examinations and treatments (Dettori, 1958).

  • Calcium determination in blood with Titriplex III and indicator buffering tablets: This study showcases the utility of Titriplex III in biochemical analysis, specifically for the precise measurement of calcium levels in blood, an essential process in various diagnostic activities (Steber, 1956).

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany
Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionHealth hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service