Skip to Content
Merck
All Photos(1)

Key Documents

1.01115

Supelco

Ammonium acetate

EMPLURA®

Synonym(s):

Ammonium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7NO2
CAS Number:
Molecular Weight:
77.08
MDL number:
UNSPSC Code:
12352107
EC Index Number:
211-162-9
NACRES:
NA.21
form:
solid
solubility:
1480 g/L

Quality Level

product line

EMPLURA®

Assay

≥96.0% (alkalimetric)

form

solid

ign. residue

≤0.02% (as sulfate)

pH

6.7-7.3 (25 °C, 50 g/L in H2O)

mp

114 °C

solubility

1480 g/L

density

1.17 g/cm3 at 20 °C

bulk density

410 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.0005%

storage temp.

15-25°C

SMILES string

[NH4]OC(=O)C

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • One-Pot, Four-Component Reaction for the Design, Synthesis, and SAR Studies of Novel Pyridines for Insecticidal Bioefficacy Screening against Cowpea Aphid (Aphis craccivora): This study explores the application of ammonium acetate in facilitating the synthesis of novel pyridine derivatives, evaluated for their insecticidal properties against Aphis craccivora (Alkorbi et al., 2024).

  • A green analytical method for the simultaneous determination of 17 perfluoroalkyl substances (PFAS) in human serum and semen by ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS): This research demonstrates the use of ammonium acetate in UPLC-MS/MS for detecting PFAS in biological samples, highlighting its role in environmental and health-related analytical chemistry (Di Giorgi et al., 2024).

  • Innovation of 6-sulfonamide-2H-chromene derivatives as antidiabetic agents targeting α-amylase, α-glycosidase, and PPAR-γ inhibitors with in silico molecular docking simulation: Ammonium acetate was utilized in the synthesis process of new chromene derivatives, showing potential as antidiabetic agents by inhibiting α-amylase and α-glycosidase activities (Thabet et al., 2024).

  • Regulation of Tetramethylpyrazine Formation by the Phenolics-Fenton Coupled Redox Cycling System: This article presents a study on the role of ammonium acetate in the formation of tetramethylpyrazine via redox cycling, an important reaction in food chemistry and possibly in synthesizing complex organic molecules (Xu et al., 2024).

  • Simultaneous determination of unecritinib (TQ-B3101) and its active metabolite crizotinib in rat plasma by LC-MS/MS: An application to pharmacokinetic studies: This paper details the use of ammonium acetate in enhancing LC-MS/MS methodologies for pharmacokinetic studies, exemplifying its critical role in the analysis of pharmaceutical compounds (Wang et al., 2024).

Analysis Note

Assay (alkalimetric): ≥ 96.0 %
Identity: passes test
pH-value (5 %; water): 6.0 - 7.5
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.0005 %
Fe (Iron): ≤ 0.001 %
Residue on ignition (as sulfate): ≤ 0.02 %
Water: ≤ 2.5 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service