Skip to Content
Merck
All Photos(2)

Key Documents

1.00984

Supelco

Isobutanol

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Isobutanol, Isobutylalcohol, 2-Methyl-1-propanol, Iso-Butanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
MDL number:
UNSPSC Code:
12191502
EC Index Number:
201-148-0
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥99% (GC)
bp:
106-108 °C/1013 hPa
vapor pressure:
12 hPa ( 20 °C)

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

12 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99% (GC)

form

liquid

autoignition temp.

430 °C

potency

>2830 mg/kg LD50, oral (Rat)
2460 mg/kg LD50, skin (Rat)

expl. lim.

1.5-12 % (v/v)

impurities

≤0.0005 meq/g Acidity
≤0.001% Peroxide (as H2O2)
≤0.01% Butyraldehyde (GC)
≤0.02% Ethylmethylketone (GC)
≤0.05% 2-Butanol (GC)
≤0.05% Isobutyraldehyde (GC)
≤0.05% Water
≤0.2% n-Butyl alcohol (GC)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n15/D 1.397-1.399

pH

7 (20 °C, 80 g/L in H2O)

bp

106-108 °C/1013 hPa

mp

-108 °C

transition temp

flash point 28 °C

solubility

85 g/L

density

0.802 g/cm3 at 20 °C

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

OCC(C)C

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service