Skip to Content
Merck
All Photos(3)

Key Documents

320048

Sigma-Aldrich

2-Methyl-1-propanol

ACS reagent, ≥99.0%

Synonym(s):

Isobutanol, Isobutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
1730878
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
108 °C (lit.)
vapor pressure:
8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

grade

ACS reagent

vapor density

2.55 (vs air)

vapor pressure

8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

801 °F

expl. lim.

10.6 %

dilution

(for analytical testing)

impurities

≤0.0005 meq/g Titr. acid
≤0.01% butyraldehyde
≤0.02% 2-butanone
≤0.1% water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.396 (lit.)

bp

108 °C (lit.)

mp

−108 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

SMILES string

CC(C)CO

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dawid Brat et al.
FEMS yeast research, 13(2), 241-244 (2013-01-03)
Simultaneous overexpression of an optimized, cytosolically localized valine biosynthesis pathway together with overexpression of xylose isomerase XylA from Clostridium phytofermentans, transaldolase Tal1 and xylulokinase Xks1 enabled recombinant Saccharomyces cerevisiae cells to complement the valine auxotrophy of ilv2,3,5 triple deletion mutants
Genny A Pang et al.
The journal of physical chemistry. A, 116(19), 4720-4725 (2012-04-21)
This work presents the first direct experimental study of the rate constant for the reaction of OH with iso-butanol (2-methyl-1-propanol) at temperatures from 907 to 1147 K at near-atmospheric pressures. OH time-histories were measured behind reflected shock waves using a
Arul M Varman et al.
Applied and environmental microbiology, 79(3), 908-914 (2012-11-28)
Global warming and decreasing fossil fuel reserves have prompted great interest in the synthesis of advanced biofuels from renewable resources. In an effort to address these concerns, we performed metabolic engineering of the cyanobacterium Synechocystis sp. strain PCC 6803 to
Jan-Karl Guterl et al.
ChemSusChem, 5(11), 2165-2172 (2012-10-23)
The limited supply of fossil resources demands the development of renewable alternatives to petroleum-based products. Here, biobased higher alcohols such as isobutanol are versatile platform molecules for the synthesis of chemical commodities and fuels. Currently, their fermentation-based production is limited
Pedram Fatehi
Biotechnology progress, 29(2), 297-310 (2013-01-09)
In this review, the recent advancements and technical challenges associated with the production of ethanol, butanol, and isobutanol via bioconversion routes from celluloses of woody materials are reviewed. Physicochemical processes, e.g. steam explosion, seem to be the most viable process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service