1.00837
Methanol
99% (GC), suitable for GC/MS, SupraSolv®
Synonym(s):
Methyl alcohol
About This Item
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Recommended Products
Product Name
Methanol, for gas chromatography MS SupraSolv®
vapor density
1.11 (vs air)
Quality Level
vapor pressure
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
product line
SupraSolv®
Assay
99% (GC)
form
liquid
autoignition temp.
725 °F
potency
5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)
expl. lim.
36 %
technique(s)
GC/MS: suitable
refractive index
n20/D 1.329 (lit.)
kinematic viscosity
0.54-0.59 cSt(20 °C)
bp
64.7 °C (lit.)
mp
−98 °C (lit.)
transition temp
flash point 9.7 °C
density
0.791 g/mL at 25 °C (lit.)
format
neat
storage temp.
2-30°C
SMILES string
CO
InChI
1S/CH4O/c1-2/h2H,1H3
InChI key
OKKJLVBELUTLKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- The composition of chemicals and anti-osteogenic properties in the volatile extracts from Homalomena gigantea rhizome: This study highlights methanol′s role in the extraction and analysis of volatile organic compounds from medicinal plants, providing insights valuable for researchers in pharmacology and natural product chemistry (Thanh et al., 2024).
- A homochiral tartrate-bridged dinuclear chromium(III) complex anion with a resonance-assisted hydrogen bond for proton conduction: Methanol is used as a solvent in the synthesis of complex inorganic compounds, which are essential in developing new materials for energy applications, such as proton exchange membranes (Dunatov et al., 2024).
Analysis Note
Other Notes
Legal Information
analytical column
solvent
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service