Skip to Content
Merck
All Photos(2)

Key Documents

1.00338

Supelco

Hydrofluoric acid

40%, for analysis EMSURE® ISO,Reag. Ph Eur

Synonym(s):

Hydrofluoric acid solution, Fluoric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

Product Name

Hydrofluoric acid 40%, for analysis EMSURE® ISO,Reag. Ph Eur

grade

reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

form

liquid

concentration

40.0-42.0% Hf

impurities

≤0.01 ppm Ba (Barium)
≤0.01 ppm Cd (Cadmium)
≤0.01 ppm Cr (Chromium)
≤0.01 ppm Li (Lithium)
≤0.02 ppm Ag (Silver)
≤0.02 ppm Be (Beryllium)
≤0.02 ppm Bi (Bismuth)
≤0.02 ppm Co (Cobalt)
≤0.02 ppm Cu (Copper)
≤0.02 ppm Ge (Germanium)
≤0.02 ppm Mo (Molybdenum)
≤0.02 ppm Ni (Nickel)
≤0.02 ppm Pb (Lead)
≤0.02 ppm Sr (Strontium)
≤0.02 ppm Ti (Titanium)
≤0.02 ppm Tl (Thallium)
≤0.02 ppm V (Vanadium)
≤0.02 ppm Zr (Zirconium)
≤0.03 ppm Arsenic and Antimony (as As)
≤0.03 ppm Mn (Manganese)
≤0.05 ppm Al (Aluminium)
≤0.05 ppm Zn (Zinc)
≤0.1 ppm Fe (Iron)
≤0.1 ppm K (Potassium)
≤0.1 ppm Mg (Magnesium)
≤0.2 ppm Ca (Calcium)
≤0.2 ppm Na (Sodium)
≤0.5 ppm Heavy metals (as Pb)
≤0.5 ppm Phosphate (PO4)
≤1 ppm Chloride (Cl)
≤5 ppm Sulfate and sulphite (as SO4)
≤50 ppm Hexafluorosilicate (SiF6)

ign. residue

≤5 ppm (as sulfate)

color

APHA: ≤10

bp

112 °C/1013 hPa

mp

-44 °C

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

Related Categories

Analysis Note

Assay (acidimetric): 40.0 - 42.0 %
Chloride (Cl): ≤ 1 ppm
Color: ≤ 10 Hazen
Hexafluorosilicate (SiF₆): ≤ 50 ppm
Phosphate (PO₄): ≤ 0.5 ppm
Sulfate and sulphite (as SO₄): ≤ 5 ppm
Arsenic and Antimony (as As): ≤ 0.03 ppm
Ag (Silver): ≤ 0.02 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 0.01 ppm
Be (Beryllium): ≤ 0.02 ppm
Bi (Bismuth): ≤ 0.02 ppm
Ca (Calcium): ≤ 0.2 ppm
Cd (Cadmium): ≤ 0.01 ppm
Co (Cobalt): ≤ 0.02 ppm
Cr (Chromium): ≤ 0.01 ppm
Cu (Copper): ≤ 0.02 ppm
Fe (Iron): ≤ 0.1 ppm
Ge (Germanium): ≤ 0.02 ppm
K (Potassium): ≤ 0.1 ppm
Li (Lithium): ≤ 0.01 ppm
Mg (Magnesium): ≤ 0.1 ppm
Mn (Manganese): ≤ 0.03 ppm
Mo (Molybdenum): ≤ 0.02 ppm
Na (Sodium): ≤ 0.2 ppm
Ni (Nickel): ≤ 0.02 ppm
Pb (Lead): ≤ 0.02 ppm
Sr (Strontium): ≤ 0.02 ppm
Ti (Titanium): ≤ 0.02 ppm
Tl (Thallium): ≤ 0.02 ppm
V (Vanadium): ≤ 0.02 ppm
Zn (Zinc): ≤ 0.05 ppm
Zr (Zirconium): ≤ 0.02 ppm
Heavy metals (as Pb): ≤ 0.5 ppm
Residue on ignition (as sulfate): ≤ 5 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service