Skip to Content
Merck
All Photos(1)

Documents

1.01834

Supelco

Hydrochloric acid

37%, for analysis EMPARTA® ACS

Synonym(s):

Hydrochloric acid 37%, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

product name

Hydrochloric acid, 37%, for analysis EMPARTA® ACS

grade

ACS reagent

Quality Level

vapor pressure

190 hPa ( 20 °C)

product line

EMPARTA®

form

liquid

concentration

36.5-38.0% HCl

impurities

≤0.01 ppm As (Arsenic)
≤0.2 ppm Fe (Iron)
≤1 ppm Free chlorine (Cl)
≤1 ppm Heavy metals (as Pb)
≤1 ppm Sulfate (SO4)
≤1 ppm Sulfite (SO3)
≤3 ppm NH4 (Ammonium)
≤50 ppm Bromide (Br)

ign. residue

≤5 ppm (as SO4)

color

APHA: ≤10

pH

<1 (20 °C in H2O)

density

1.19 g/cm3 at 20 °C

storage temp.

15-25°C

General description

for analysis EMPARTA® ACS

Application


  • Speciation of tin ions in oxide glass containing iron oxide through solvent extraction and inductively coupled plasma atomic emission spectrometry after the decomposition utilizing ascorbic acid: This research employs hydrochloric acid as a critical component in the dissolution process, facilitating advanced analytical techniques for material analysis. The study is pivotal for developing new methodologies in the recycling and efficient use of materials (Saijo et al., 2022).

  • Simple Preparation of Carbon Dots and Application in Cephalosporin Detection: Highlighting the use of hydrochloric acid in synthesizing carbon dots for cephalosporin detection, this paper introduces a novel approach to enhancing the sensitivity and specificity of antibiotic detection in complex samples, proving essential for both environmental monitoring and pharmaceutical compliance (Li et al., 2021).

  • Assessing the behaviour of heavy metals in abandoned phosphogypsum deposits combining electrical resistivity tomography and multivariate analysis: This study utilizes hydrochloric acid in geochemical assays to assess pollutant distribution and mobility. It offers significant insights into environmental safety and pollution control strategies, particularly in the management of industrial waste residues (Vásconez-Maza et al., 2021).

  • Fructan from Polygonatum cyrtonema Hua as an eco-friendly corrosion inhibitor for mild steel in HCl media: This investigation explores the use of natural fructan in the presence of hydrochloric acid to inhibit corrosion, providing an environmentally friendly alternative to traditional corrosion inhibitors. Such studies are crucial for sustainable industrial practices (Zhang et al., 2020).


  • Monitoring the extraction of copper from chicken dung leachate using an aluminium electrode as an indicator: Hydrochloric acid plays a vital role in this novel electrochemical method to monitor copper levels in agricultural waste products, showcasing its importance in agricultural waste management and resource recovery (Kugeria et al., 2019).

Analysis Note

Assay (alkalimetric)36.5 - 38.0 %

Identity passes test

Appearance passes test

Color≤ 10 Hazen

Residue on ignition (as SO₄)≤ 5 ppm

Expiry date: see product label


Conforms to the ACS specification for general use

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service