Skip to Content
Merck
All Photos(2)

Documents

1.00334

Supelco

Hydrofluoric acid

48%, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Hydrofluoric acid solution, Fluoric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

product name

Hydrofluoric acid 48%, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

form

liquid

pH

2 (20 °C in H2O)

mp

-35 °C

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

General description

Hydrofluoric acid (EMSURE® grade product) is used for regulated analyses and highly demanding lab applications. It also offers the highest quality and unmatched scope of specifications to give complete control of test conditions and eliminate uncertainties. EMSURE® grade products comply with all international regulations and can be used in various applications.

Application

Hydrofluoric acid 48% can be used as:
  • An etchant for the electrochemical wet etching of single-crystalline silicon wafers.
  • A matrix for the surface patterning of silicon via metal deposition.
  • A digestive acid for the determination of lanthanides in soils and sediments by inductively coupled plasma mass spectrometry.

Features and Benefits

  • Most extensive specifications worldwide
  • Tested for up to 70 parameters
  • Extraordinary purity
  • Verylow limiting values
  • Greater accuracy and control of analyses
  • Optimized for highly critical and demanding analyses
  • Ideal for method development
  • No interference or contamination due to unknown impurities

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service