Skip to Content
Merck
All Photos(1)

Documents

P-903

Supelco

Protriptyline hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N · HCl
CAS Number:
Molecular Weight:
299.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CNCCCC1C2=C(C=CC=C2)C=CC3=C1C=CC=C3.[H]Cl

InChI

1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-10,12-13,19-20H,6,11,14H2,1H3;1H

InChI key

OGQDIIKRQRZXJH-UHFFFAOYSA-N

Gene Information

General description

Protriptyline is a second generation tricyclic antidepressant (TCA) marketed under the trade name Vivactil®, for treatment of major depression and ADHD. This certified Snap-N-Spike® solution is applicable for use in LC/MS or GC/MS applications in clinical toxicology, forensic analysis, or urine drug testing. Protriptyline is unique among TCAs, since it has energizing, rather than sedating effects.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vivactil is a registered trademark of Teva Women's Health, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C C Lin
Journal of the Formosan Medical Association = Taiwan yi zhi, 92 Suppl 4, S232-S236 (1993-12-01)
To determine the effects of protriptyline, a tricyclic non-sedative antidepressant, on sleep architecture, day and night arterial oxygen saturation and pulmonary function, ten patients with stable chronic obstructive pulmonary disease were evaluated. Overnight polysomnographic measurements were made before and 2
R E Dahl et al.
Journal of the American Academy of Child and Adolescent Psychiatry, 33(6), 834-841 (1994-07-01)
Although narcolepsy is rarely diagnosed before adulthood, symptoms often begin much earlier and can easily mimic psychiatric disorders in children and adolescents. Clinical experience from a pediatric sleep center is reviewed in 16 consecutive cases of polysomnographically proven narcolepsy with
G M Barthlen
Respiration; international review of thoracic diseases, 64 Suppl 1, 35-38 (1997-01-01)
Patients with neuromuscular disease may suffer from nocturnal respiratory failure despite normal daytime respiratory function. The physiological reduction in muscle tone during sleep may be life-threatening in a patient with impaired muscle strength. Nocturnal respiratory failure may occur in patients
P Lévy et al.
Sleep, 19(9 Suppl), S101-S110 (1996-11-01)
The spectrum of respiratory sleep disorders has been extended in the last years to include conditions that are less well defined than severe obstructive sleep apnea (OSA). Moderate OSA, snoring, and upper airway resistance syndrome (UARS) represent three conditions in
Y Oku et al.
Respiration physiology, 93(1), 41-49 (1993-07-01)
Protriptyline (PRT) has been shown to preferentially stimulate upper airway inspiratory motorneurons relative to phrenic activity in hyperoxic hypercapnia in the decerebrate cat via a carotid body-independent mechanism. Since previous studies indicated that carotid body stimulation results in preferential activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service