Skip to Content
Merck
All Photos(1)

Key Documents

D-906

Supelco

Desipramine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23ClN2
CAS Number:
Molecular Weight:
302.84
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.CNCCCN1c2ccccc2CCc3ccccc13

InChI

1S/C18H22N2.ClH/c1-19-13-6-14-20-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)20;/h2-5,7-10,19H,6,11-14H2,1H3;1H

InChI key

XAEWZDYWZHIUCT-UHFFFAOYSA-N

Gene Information

human ... SLC6A2(6530)

General description

A Certified Solution Standard suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing. Desipramine is a tricyclic antidepressant (TCA) sold under the brand names Norpramin® and Pertofrane. This TCA drug is also a primary active metabolite of imipramine.

Application


  • Pharmacological Treatments for Localized Provoked Vulvodynia: Reviews current pharmacological treatments, including tricyclic antidepressants like desipramine hydrochloride, emphasizing their role in managing chronic pain conditions, crucial for ongoing research in neuropharmacology and clinical practice (Bajzak et al., 2023).

  • Desipramine induces anti-inflammatory dorsal root ganglion transcriptional signatures: Highlights the use of desipramine hydrochloride in neurobiological research, specifically its effects on inflammatory responses in nerve injury models, underscoring its significance in neuropharmacological studies (Serafini et al., 2024).

  • Effect of lipid emulsion on neuropsychiatric drug-induced toxicity: Discusses the therapeutic potential of lipid emulsions in managing toxicity caused by drugs, including desipramine hydrochloride, which is essential for improving safety profiles in clinical settings (Hwang and Sohn, 2024).

  • Sustainable beeswax modified cellulose paper for the determination of tricyclic antidepressants: Researches innovative methods for detecting tricyclic antidepressants like desipramine hydrochloride in biofluids, pivotal for advancing forensic and clinical toxicology (González-Bermúdez et al., 2024).

  • Sleep-wake dependent hippocampal regulation of fear memory: Although indirectly related, this study forms part of the broader context of neurological research where desipramine hydrochloride could be used to modulate neurochemical pathways, relevant for understanding sleep-related memory processes (Wen et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Norpramin is a registered trademark of Aventisub II Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Richard M O'Connor et al.
Pharmacology, biochemistry, and behavior, 103(3), 561-567 (2012-10-13)
The neurotransmitter glutamate is increasingly being implicated as playing a role in the molecular pathology underlying depression. The group III family of metabotropic glutamate (mGlu) receptors (mGlu(4,) mGlu(7) and mGlu(8) receptors) remains the most poorly investigated of all glutamate receptors
O F O'Leary et al.
Neuroscience, 228, 36-46 (2012-10-17)
Approximately 50% of patients with a major depressive episode fail to achieve remission with first-line antidepressant treatments. Second-line treatment strategies for such patients include lithium augmentation of antidepressants, particularly with tricyclic antidepressants. The neurobiological mechanisms underlying the therapeutic effects of
Alain Watts et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(39), 13411-13420 (2012-09-28)
Rapid eye movement (REM) sleep enhances hippocampus-dependent associative memory, but REM deprivation has little impact on striatum-dependent procedural learning. Antidepressant medications are known to inhibit REM sleep, but it is not well understood if antidepressant treatments impact learning and memory.
Thomas M Polasek et al.
British journal of clinical pharmacology, 75(4), 1019-1028 (2012-08-29)
To determine the effect of increasing adult age on predicted metabolic drug clearance. Predicted metabolic drug clearances (CLPT ) were determined using in vitro-in vivo extrapolation coupled with physiological-based pharmacokinetic modelling and simulation (IVIVE-PBPK) in Simcyp®. Simulations were conducted using
Maria Stratinaki et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(20), 8254-8259 (2013-05-01)
Regulator of G protein signaling 4 (Rgs4) is a signal transduction protein that controls the function of monoamine, opiate, muscarinic, and other G protein-coupled receptors via interactions with Gα subunits. Rgs4 is expressed in several brain regions involved in mood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service