Skip to Content
Merck
All Photos(1)

Key Documents

F-903

Supelco

Fluphenazine dihydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N3OSF3 · H2Cl2
CAS Number:
Molecular Weight:
510.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

FC(F)(F)c1cc2c(cc1)Sc3c(cccc3)N2CCC[N+H]4CC[N+H](CC4)CCO.[Cl-].[Cl-]

InChI

1S/C22H26F3N3OS.2ClH/c23-22(24,25)17-6-7-21-19(16-17)28(18-4-1-2-5-20(18)30-21)9-3-8-26-10-12-27(13-11-26)14-15-29;;/h1-2,4-7,16,29H,3,8-15H2;2*1H

InChI key

MBHNWCYEGXQEIT-UHFFFAOYSA-N

Gene Information

human ... DRD2(1813)

General description

Fluphenazine, sold under the trade name Prolixin®, is a typical antipsychotic prescribed for treatment of schizophrenia, mania, bipolar disorder and dementia. This certified solution standard is suitable for fluphenazine testing methods by GC/MS or LC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.

Application


  • Monitoring Fluphenazine Metabolites in Schizophrenia: A study developed a radioimmunoassay for the 7-hydroxy metabolite of fluphenazine, applied to monitor plasma levels in patients treated long-term with oral and depot fluphenazine. This research enhances understanding of fluphenazine′s pharmacokinetics and aids in optimizing treatment regimens for schizophrenia, contributing to personalized medicine approaches (Aravagiri et al., 1994).

  • Neuroleptics in Clinical Practice: A review discusses the development and application of assays for neuroleptics, including fluphenazine, providing clinical perspectives on their use. This research is vital for enhancing therapeutic strategies and understanding the pharmacological profiles of antipsychotic drugs, which are crucial in treating severe mental disorders (Midha et al., 1993).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Prolixin is a registered trademark of E.R. Squibb & Sons, LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alcindo Busanello et al.
Pharmacology, biochemistry, and behavior, 101(2), 307-310 (2012-01-24)
Treatment with classical neuroleptics in humans can produce a serious side effect, known as tardive dyskinesia (TD). Here, we examined the possible neuroprotective effects of resveratrol, a polyphenol compound contained in red grapes and red wine, in an animal model
Yu-Wen Chen et al.
European journal of pharmacology, 693(1-3), 31-36 (2012-08-29)
The aim of this study was to examine whether thioxanthine-type antipsychotics (chlorprothixene and cis(z)-flupenthixol) and phenothiazine-type antipsychotics (chlorpromazine and fluphenazine) produced spinal anesthesia. Using a rat model of intrathecal injection, we evaluated spinal anesthesia of antipsychotic drugs (chlorprothixene, cis(z)-flupenthixol, chlorpromazine
Martín Klappenbach et al.
Learning & memory (Cold Spring Harbor, N.Y.), 19(2), 73-83 (2012-01-24)
The understanding of how the reinforcement is represented in the central nervous system during memory formation is a current issue in neurobiology. Several studies in insects provide evidence of the instructive role of biogenic amines during the learning and memory
David G Goodwin et al.
The Journal of parasitology, 97(1), 148-151 (2011-02-26)
An increasing interest in the association of the presence of antibodies to Toxoplasma gondii and the development of schizophrenia in patients has been generated over the last several years. Some antischizophrenia agents have been shown to have activity against T.
Thomas E Wooters et al.
Brain research, 1396, 45-53 (2011-05-17)
Impulsivity is one of the core symptoms of attention-deficit/hyperactivity disorder (ADHD). The spontaneously hypertensive rat (SHR), a putative animal model of ADHD, has been used to investigate the neurobiology of impulsivity, although this model has been questioned over concerns that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service