Skip to Content
Merck
All Photos(1)

Key Documents

P-051

Supelco

PMMA hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

p-Methoxymethamphetamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18ClNO
CAS Number:
Molecular Weight:
215.72
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CNC(C)Cc1ccc(OC)cc1

InChI

1S/C11H17NO.ClH/c1-9(12-2)8-10-4-6-11(13-3)7-5-10;/h4-7,9,12H,8H2,1-3H3;1H

InChI key

IQZVXWOBOYTPER-UHFFFAOYSA-N

General description

PMMA, or para-methoxymethamphetamine, is a stimulant and psychedelic drug closely related to the amphetamine PMA. This designer drug is sometimes used as a substitute for the amphetamine MDMA. This Certified Spiking Solution® is suitable for use in LC/MS or GC/MS amphetamine testing methods in clinical toxicology, forensic analysis, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T D Steele et al.
Brain research, 589(2), 349-352 (1992-09-04)
These studies assessed the neurotoxic potential of N-methyl-1-(4-methoxyphenyl)-2-aminopropane (para-methoxymethamphetamine; PMMA), an amphetamine analog that has surfaced in the illicit drug market. Repeated subcutaneous injections of PMMA caused lasting, dose-related reductions in regional brain concentrations of serotonin (5-HT) and 5-hydroxyindoleacetic acid
Roland F Staack et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(4), 379-381 (2004-03-25)
p-Methoxymethamphetamine (PMMA) is a new designer drug, listed in many countries as a controlled substance. Several fatalities have been attributed to the abuse of this designer drug. Previous in vivo studies using Wistar rats had shown that PMMA was metabolized
R A Glennon et al.
Pharmacology, biochemistry, and behavior, 31(1), 9-13 (1988-09-01)
The controlled-substance analog N-monomethyl-1-(4-methoxyphenyl)-2-aminopropane (PMMA) may be viewed as being either the 4-methoxy analog of methamphetamine or the N-methyl analog of 1-(4-methoxyphenyl)-2-aminopropane (PMA). Because of its abuse potential, PMMA was examined with regard to (a) its stimulus properties in rats
Tomáš Páleníček et al.
Pharmacology, biochemistry, and behavior, 98(1), 130-139 (2010-12-21)
Despite poisoning with the ecstasy substitute para-methoxymethamphetamine (PMMA) being typically associated with severe hyperthermia and death, behavioral and toxicological data on this drug are missing. Herein we present the behavioral profile of PMMA, its hyperthermic potency and pharmacokinetic profile in
Roland F Staack et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 789(1), 27-41 (2003-05-03)
Studies are described on the metabolism and the toxicological analysis of the new designer drug rac-p-methoxymethamphetamine (PMMA) in rat urine using gas chromatography-mass spectrometry (GC-MS). The identified metabolites indicated that PMMA was extensively metabolized mainly by O-demethylation to pholedrine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service