Skip to Content
Merck
All Photos(1)

Key Documents

M-020

Supelco

S(+)-Methamphetamine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

dextro-Methamphetamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CN[C@@H](C)Cc1ccccc1

InChI

1S/C10H15N/c1-9(11-2)8-10-6-4-3-5-7-10/h3-7,9,11H,8H2,1-2H3/t9-/m0/s1

InChI key

MYWUZJCMWCOHBA-VIFPVBQESA-N

Gene Information

General description

Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes. Known by the street names "meth," "ice," and "glass", this Snap-N-Spike® reference solution is applicable for use in LC/MS or GC/MS applications including clinical toxicology, sports testing, forensic analysis, or urine drug testing. The dextrorotatory isomer of methamphetamine is sold under the trade name Desoxyn® for treatment of ADHD and obesity.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Desoxyn is a registered trademark of Abbott Laboratories Corp.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Kelly
Emergency medicine journal : EMJ, 30(4), 339-340 (2013-03-21)
A short-cut review was carried out to establish which clinical parameters are useful in predicting outcome in patients who present having body packed metamphetamine. Eleven papers were found using the reported searches, of which one presented the best evidence to
Diane M Herbeck et al.
Journal of psychoactive drugs, 45(2), 132-140 (2013-08-03)
This study examines health and legal problems associated with use of commonly reported substances and combinations of substances in a sample of adults with long histories of methamphetamine (meth) use. Data are from a 2009-11 eight-year follow-up interview in an
New Meth vaccine shows promise in animals.
Human vaccines & immunotherapeutics, 9(1), 9-10 (2013-04-10)
Jeffrey T Parsons et al.
AIDS and behavior, 17(4), 1478-1487 (2013-04-05)
Methamphetamine use is associated with HIV infection, especially among gay and bisexual men. Methamphetamine use contributes to disease progression both directly, by increasing viral load and damaging the immune system, and indirectly, by decreasing medication adherence. Research examining the association
Glen R Hanson et al.
The Journal of pharmacology and experimental therapeutics, 346(2), 173-181 (2013-05-21)
Because of persistent social problems caused by methamphetamine (METH), new therapeutic strategies need to be developed. Thus, we investigated the response of central nervous system neurotensin (NT) systems to METH self-administration (SA) and their interaction with basal ganglia dopamine (DA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service