Skip to Content
Merck
All Photos(1)

Key Documents

P-050

Supelco

PMA hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

p-Methoxyamphetamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16ClNO
CAS Number:
Molecular Weight:
201.69
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

[Cl-].NC(Cc1ccc(cc1)OC)C.[H+]

InChI

1S/C10H15NO.ClH/c1-8(11)7-9-3-5-10(12-2)6-4-9;/h3-6,8H,7,11H2,1-2H3;1H

InChI key

VSHZXOLHFRVZND-UHFFFAOYSA-N

General description

PMA, or para-methoxyamphetamine is a ring-substituted phenethylamine with hallucinogenic effects. PMA is known by street names "Dr. Death," "Chicken Powder," and "Chicken Yellow." Toxicity of PMA may occur at doses only slightly above the usual recreation dose.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul D Callaghan et al.
Journal of neurochemistry, 100(3), 617-627 (2006-12-22)
p-Methoxyamphetamine (PMA) has been implicated in fatalities as a result of 'ecstasy' (MDMA) overdose worldwide. Like MDMA, acute effects are associated with marked changes in serotonergic neurotransmission, but the long-term effects of PMA are poorly understood. The aim of this
M M W Straiko et al.
Neuroscience, 144(1), 223-231 (2006-11-07)
The present study quantified the cleaved form of the microtubule-associated protein tau (cleaved MAP-tau, C-tau), a previously demonstrated marker of CNS toxicity, following the administration of monoamine-depleting regimens of the psychostimulant drugs amphetamine (AMPH), methamphetamine (METH), +/-3,4-methylenedioxymethamphetamine (MDMA), or para-methoxyamphetamine
Amparo Fornés et al.
The Biochemical journal, 412(3), 495-506 (2008-03-18)
The neuronal glycine transporter GLYT2 controls the availability of the neurotransmitter in glycinergic synapses, and the modulation of its function may influence synaptic transmission. The active transporter is located in membrane rafts and reaches the cell surface through intracellular trafficking.
Fatal paramethoxy-amphetamine (PMA) poisoning in the Australian Capital Territory.
Paul G Lamberth et al.
The Medical journal of Australia, 188(7), 426-426 (2008-04-09)
Diego Bustamante et al.
Pharmacology, biochemistry, and behavior, 79(2), 199-212 (2004-10-27)
The analgesic effects of (+)- and (-)-amphetamine (AMPH), (+/-)-p-methoxyamphetamine (MA), (+/-)-N-methyl-p-methoxyamphetamine (MMA) and (+/-)-N-ethyl-p-methoxyamphetamine (EMA) were compared using two different algesimetric tests in rats. In the formalin test, (+)-AMPH elicited significant antinociception at doses of 0.2, 2 and 8 mg/kg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service