Skip to Content
Merck
All Photos(1)

Key Documents

81489

Supelco

Poly(methyl methacrylate)

analytical standard, for GPC, 2,000

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard
for GPC

Quality Level

autoignition temp.

580 °F

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~1730
Mp ~1960
Mw ~1900

technique(s)

gel permeation chromatography (GPC): suitable

solubility

H2O: insoluble

density

1.200 g/cm3

Mw/Mn

~1.10

format

neat

storage temp.

2-8°C

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(methyl methacrylate) is a rigid and transparent type of polymer material.[1] It is obtained via polymerization of methyl methacrylate monomer.[2]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plastics Materials (1999)
Biomechanics and Biomaterials in Orthopedics (2004)
Kariem Mostafa Al Batouty et al.
The journal of adhesive dentistry, 15(2), 161-166 (2013-03-28)
To compare the push-out bond strength of a new glass fiber root canal filling material to that of Resilon/ Epiphany, EndoREZ, and gutta-percha/AH Plus root canal filling materials. Sixty roots were randomly divided into four groups (n = 15) according
S A Bini et al.
The bone & joint journal, 95-B(3), 367-370 (2013-03-02)
We evaluated the impact of pre-coating the tibial component with polymethylmethacrylate (PMMA) on implant survival in a cohort of 16 548 primary NexGen total knee replacements (TKRs) in 14 113 patients. In 13 835 TKRs a pre-coated tray was used
Distortions of iris and anterior lens capsule OCT images by ICRS.
Hosik Hwang et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(3), 155-156 (2013-03-01)

Questions

  1. What are the alpha- and omega-ends of this PMMA?

    1 answer
    1. Unfortunately, there is no information on the alpha and omega ends for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service