Skip to Content
Merck
All Photos(1)

Key Documents

79346

Sigma-Aldrich

Phorbol 12-myristate 13-acetate

synthetic, ≥98.0% (TLC)

Synonym(s):

12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H56O8
CAS Number:
Molecular Weight:
616.83
Beilstein:
2407201
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (TLC)

form

film

quality

synthetic

solubility

chloroform: 10 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)C(C=C(CO)C[C@@]3(O)C2C=C(C)C3=O)C4C(C)(C)[C@]14OC(C)=O

InChI

1S/C36H56O8/c1-7-8-9-10-11-12-13-14-15-16-17-18-29(39)43-32-24(3)35(42)27(30-33(5,6)36(30,32)44-25(4)38)20-26(22-37)21-34(41)28(35)19-23(2)31(34)40/h19-20,24,27-28,30,32,37,41-42H,7-18,21-22H2,1-6H3/t24-,27+,28-,30-,32-,34-,35-,36-/m1/s1

InChI key

PHEDXBVPIONUQT-RGYGYFBISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Phorbol 12-myristate 13-acetate has been used to stimulate the in vitro production of interleukin (IL)-18, IL-10[1] and macrophages.[2] It has also been used to induce protein kinase C activity.[3]

Biochem/physiol Actions

Phorbol 12-myristate 13-acetate is a potent tumor promoter and activates protein kinase C in vivo and in vitro. It is a phorbol ester that is associated with many cellular responses including gene transcription, cell division and differentiation, apoptosis and immune response. It is also involved in protein kinase C (PKC) mediated receptor desensitization.[4] Phorbol 12-myristate 13-acetate induces PKC localization from the cytosol,[5] enabling the enzyme association with the cell membrane.[6]

Other Notes

Activates protein kinase C in vivo and in vitro at nanomolar concentrations[7]; Highly potent tumor promotor and mitogen: active principle of croton oil. Review[8]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Kontny et al.
Eur. J. Pharmacol., Molec. Pharmacol. Sec., 227, 333-333 (1992)
The calcium ATPase SERCA2 regulates desmoplakin dynamics and intercellular adhesive strength through modulation of PKCalpha signaling
Hobbs RP, et al.
Faseb Journal, 25(3), 990-1001 (2011)
R D Estensen
Journal of experimental pathology, 1(2), 71-77 (1984-01-01)
Tumor promoters act on carcinogen-initiated tissues to cause phenotypic expression of malignancy. Phorbol ester tumor promoters, like hormones, act on cells and tissues at nanomolar concentrations, often producing "physiological" effects. These promoters also act on cells to produce what appear
Differential effect of bryostatin 1 and phorbol 12-myristate 13-acetate on HOP-92 cell proliferation is mediated by down-regulation of protein kinase Cdelta
Choi SH, et al.
Cancer Research, 66(14), 7261-7269 (2006)
Adipose tissue-derived mesenchymal stem cells are more potent suppressors of dendritic cells differentiation compared to bone marrow-derived mesenchymal stem cells
Ivanova-Todorova E, et al.
Immunology Letters, 126(1-2), 37-42 (2009)

Questions

1–6 of 6 Questions  
  1. How many ML of DMSO should I use to dilute each milligram of phorbol?

    1 answer
    1. This product can be solubilized in DMSO at 10 mg/mL.

      Helpful?

  2. How should Product 79346, Phorbol 12-myristate 13-acetate - synthetic, be dissolved?

    1 answer
    1. PMA is soluble in acetone, DMSO, ethyl acetate, ethanol and methylene chloride, but is practically insoluble in water. The solubility of 79346 has been tested in chloroform at a concentration of 10 mg/mL.  Stock solutions of 20 mM can be prepared in DMSO and diluted into aqueous buffers.

      Helpful?

  3. How can I store the Product 79346, Phorbol 12-myristate 13-acetate - synthetic (PMA), solution solution?  How long?

    1 answer
    1. Stock solutions in DMSO can be stored protected from light as aliquots at -20°C for at least 6 months, or at 2-8°C for up to 3 months.

      Helpful?

  4. Why do I not see any solid in the bottle of Product 79346, Phorbol 12-myristate 13-acetate - synthetic?

    1 answer
    1. Phorbol 12-myristate 13-acetate may be an invisible film in the bottle.

      Helpful?

  5. What is the difference between product 79346, Phorbol 12-myristate 13-acetate - synthetic, and molecular biology grade product P1585?

    1 answer
    1. Product P1585 has been tested and found suitable for stimulation of interleukin 2 production of Jurkat cells. Product 79346 has not tested in this application.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service