Skip to Content
Merck
All Photos(1)

Key Documents

F-017

Supelco

(±)-Flecainide solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20F6N2O3
CAS Number:
Molecular Weight:
414.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(NCC1CCCCN1)C2=CC(OCC(F)(F)F)=CC=C2OCC(F)(F)F

InChI

1S/C17H20F6N2O3/c18-16(19,20)9-27-12-4-5-14(28-10-17(21,22)23)13(7-12)15(26)25-8-11-3-1-2-6-24-11/h4-5,7,11,24H,1-3,6,8-10H2,(H,25,26)

InChI key

DJBNUMBKLMJRSA-UHFFFAOYSA-N

Gene Information

human ... SCN5A(6331)

General description

Flecainide, marketed under trade names such as Almarytm, Apocard, Ecrinal®, and Flécaine, is a class Ic antiarrhythmic drug used to prevent and treat a variety of cardiac arrhythmias. This Certified Spiking Solution® is suitable for use as a starting material in the preparation of calibrators, controls, or linearity standards in flecainide therapeutic drug monitoring methods by LC/MS or LC that ensure patients remain within the drug′s narrow therapeutic range.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Ecrinal is a registered trademark of Laboratoires Asepta Corporation Monaco
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

ECG diagnosis: flecainide toxicity.
Joel T Levis
The Permanente journal, 16(4), 53-53 (2012-12-20)
Joseph J Lynch et al.
Journal of cardiovascular pharmacology, 61(3), 226-232 (2012-11-29)
This study explored the intrinsic vasorelaxant and inotropic effects of the mixed potassium and sodium channel blocker atrial antiarrhythmic vernakalant and the class IC antiarrhythmic agent flecainide in human isolated subcutaneous resistance artery and in ventricular trabecular muscle preparations. At
David H Jang et al.
The Journal of emergency medicine, 44(4), 781-783 (2012-09-18)
Flecainide is a class IC antidysrhythmic primarily indicated for ventricular dysrhythmias and supraventricular tachycardia (SVT). Class IC antidysrhythmic overdose has a reported mortality of 22%, and death results from dysrhythmias and cardiovascular collapse. We report a near-fatal flecainide overdose in
Antiarrhythmic drug treatment after cardioversion of atrial fibrillation.
Marcelle D Smit et al.
Lancet (London, England), 380(9852), 1467-1467 (2012-10-30)
Kosuke Doki et al.
Pharmacogenetics and genomics, 22(11), 777-783 (2012-09-04)
To investigate the association between age-related decline in flecainide clearance and CYP2D6 genotype, we conducted a population pharmacokinetic analysis of flecainide using routine therapeutic drug monitoring data. Population pharmacokinetic analysis was performed on retrospective data from 163 genotyped patients treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service