Skip to Content
Merck
All Photos(1)

Key Documents

H-108

Supelco

trans-3′-Hydroxycotinine-D3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9D3N2O2
CAS Number:
Molecular Weight:
195.23
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C10H12N2O2/c1-12-8(5-9(13)10(12)14)7-3-2-4-11-6-7/h2-4,6,8-9,13H,5H2,1H3/t8-,9+/m0/s1/i1D3

InChI key

XOKCJXZZNAUIQN-IRPRQUBLSA-N

General description

Trans-3′-Hydroxycotinine is a major nicotine metabolite excreted in the urine of smokers and users of other tobacco or nicotine products including cigars, chewing tobacco and snuff. This certified Snap-N-Spike® solution is suitable for use as an internal standard for quantification of trans-3′-Hydroxycotinine levels in urine or other matrices for nicotine testing by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Bertuzzi et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(42), E9926-E9933 (2018-10-03)
A particularly essential determinant of a neuron's functionality is its neurotransmitter phenotype. While the prevailing view is that neurotransmitter phenotypes are fixed and determined early during development, a growing body of evidence suggests that neurons retain the ability to switch
Balmiki Ray et al.
Current protocols in neuroscience, Chapter 7, Unit 7-Unit 7 (2012-07-04)
Acetylcholine (ACh) is the neurotransmitter used by cholinergic neurons at the neuromuscular junction, in parasympathetic peripheral nerve terminals, and in important memory-related circuits in the brain, and takes part in other critical functions. ACh is synthesized from choline and acetyl
Julie Routhier et al.
Nature communications, 12(1), 6384-6384 (2021-11-06)
Chronic Obstructive Pulmonary Disease is a generally smoking-linked major cause of morbidity and mortality. Genome-wide Association Studies identified a locus including a non-synonymous single nucleotide polymorphism in CHRNA5, rs16969968, encoding the nicotinic acetylcholine receptor α5 subunit, predisposing to both smoking
Balmiki Ray et al.
Brain research, 1297, 160-168 (2009-08-08)
Cholinergic neurons are a major constituent of the mammalian central nervous system. Acetylcholine, the neurotransmitter used by cholinergic neurons, is synthesized from choline and acetyl CoA by the enzymatic action of choline acetyltransferase (ChAT). The transport of choline into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service