Y102
YC-1
powder
Synonym(s):
3-(5′-Hydroxymethyl-2′-furyl)-1-benzyl indazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H16N2O2
CAS Number:
Molecular Weight:
304.34
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
form
powder
color
white to yellow
solubility
DMSO: 10 mg/mL
H2O: insoluble
SMILES string
OCc1ccc(o1)-c2nn(Cc3ccccc3)c4ccccc24
InChI
1S/C19H16N2O2/c22-13-15-10-11-18(23-15)19-16-8-4-5-9-17(16)21(20-19)12-14-6-2-1-3-7-14/h1-11,22H,12-13H2
InChI key
OQQVFCKUDYMWGV-UHFFFAOYSA-N
Gene Information
human ... PDE5A(8654)
Application
YC-1 has been used as a hypoxia-inducible factor 1α (HIF-1α) inhibitor:
- to reduce hypoxia induced Jagged1 expression in cardiomyocytes (CMs)
- to study its effect on progenitor expansion and CD34+ and side population (SP) cell phenotype and on the proliferation rate of cells with an ability to form long term colony forming units
- to study its effect on regulating sphingosine 1-phosphate (S1P) bound to albumin induced plasminogen activator inhibitor 1 (PAI-1) expression by activating Rho/ Rho-associated protein kinase (ROCK) pathway.
Biochem/physiol Actions
NO (nitric oxide)-independent activator of soluble guanylyl cyclase.
YC-1 activates soluble guanylyl cyclase and prevents platelet aggregation and vascular contraction. It has a potential to treat circulation disorders. YC-1 also has an ability to inhibit hypoxia-inducible factor 1α (HIF-1α) activity in vitro. It acts as a potential antiangiogenic anticancer agent.
Features and Benefits
This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hypoxia-stressed cardiomyocytes promote early cardiac differentiation of cardiac stem cells through HIF-1$\alpha$/Jagged1/Notch1 signaling
Wang K, et al.
Acta Pharmaceutica Sinica. B, 8(5), 795-804 (2018)
YC-1: a potential anticancer drug targeting hypoxia-inducible factor 1
Yeo EJ, et al.
Journal of the National Cancer Institute, 95(7), 516-525 (2003)
Zong-Tao Chai et al.
PloS one, 8(10), e77957-e77957 (2013-11-07)
microRNAs (miRNAs) have been reported to regulate angiogenesis by down-regulating the expression of pro-angiogenic or anti-angiogenic factors. The aims of this study were to investigate whether miR-26a inhibited angiogenesis by down-regulating vascular endothelial growth factor A (VEGFA) and its clinical
Vehicle-dependent effects of sphingosine 1-phosphate on plasminogen activator inhibitor-1 expression
Takahashi C, et al.
Journal of Atherosclerosis and Thrombosis, 37663-37663 (2017)
F N Ko et al.
Blood, 84(12), 4226-4233 (1994-12-15)
YC-1 [3-(5'-hydroxymethyl-2'-furyl)-1-benzylindazole] inhibited the aggregation of and ATP release from washed rabbit platelets induced by arachidonic acid (AA), collagen, U46619, platelet-activating factor (PAF), and thrombin in a concentration-dependent manner. YC-1 also disaggregated the clumped platelets caused by these inducers. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service