Skip to Content
Merck
All Photos(3)

Key Documents

840101P

Avanti

Egg PA

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidic acid (Egg, Chicken) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840101P-1g)
pkg of 2 × 100 mg (840101P-200mg)
pkg of 5 × 100 mg (840101P-500mg)
pkg of 1 × 25 mg (840101P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H71O8P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(39)45-35(34-44-46(40,41)42)33-43-36(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,35H,3-16,19-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/b18-17-;/t35-;/m1./s1

InChI key

PPYXMAJBOKWTQX-RYRMQHSSSA-M

General description

Phosphatidic acid is a simple and basic glycerophospholipid. It is an ester of glycerol with two long chain fatty acids at C1 and C2 position and phosphoric acid at C3 position. PA is synthesized by the hydrolytic action of phospholipase D on glycerophospholipids.

Application

Egg PA is suitable for use:
  • in the preparation of reconstituted high density lipoprotein (rHDL) along with serum amyloid A (SAA)
  • as negatively charged lipid in the preparation of giant unilamellar vesicles
  • in the preparation of liposomes

Biochem/physiol Actions

Phosphatidic acid (PA) is involved in the synthesis of phosphatidylcholine, phosphatidylethanolamine, phosphatidylinositol, phosphatidylserine and cardiolipin. PA functions as a signaling molecule.

Packaging

5 mL Clear Glass Sealed Ampule (840101P-200mg)
5 mL Clear Glass Sealed Ampule (840101P-25mg)
5 mL Clear Glass Sealed Ampule (840101P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (840101P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemistry of Lipids, Lipoproteins and Membranes (1996)
Hepatitis B virus X protein induces size-selective membrane permeabilization through interaction with cardiolipin
You DG, et al.
Biochimica et Biophysica Acta - Biomembranes, 1861(4), 729-737 (2019)
Effects of lipid composition on the structural properties of human serum amyloid A in reconstituted high-density lipoprotein particles
Takase H, et al.
Chemistry and Physics of Lipids, 221, 8-14 (2019)
Muscle and Exercise Physiology (2018)
Membrane permeabilization by trypanosome lytic factor, a cytolytic human high density lipoprotein
Harrington JM, et al.
The Journal of Biological Chemistry, 284(20), 13505-13512 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service