Skip to Content
Merck
All Photos(3)

Key Documents

840042P

Avanti

Liver PI

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylinositol (Liver, Bovine) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840042P-10mg)
pkg of 1 × 100 mg (840042P-100mg)
pkg of 1 × 25 mg (840042P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]([C@H](O)[C@H](O)[C@@H](O)[C@@H]1O)OP([O-])(OC[C@@]([H])(COC(CCCCCCCCCCCCCCCCC)=O)OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)=O.[Na+]

InChI

1S/C47H83O13P.Na/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-41(49)59-39(38-58-61(55,56)60-47-45(53)43(51)42(50)44(52)46(47)54)37-57-40(48)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2;/h11,13,17,19,22,24,28,30,39,42-47,50-54H,3-10,12,14-16,18,20-21,23,25-27,29,31-38H2,1-2H3,(H,55,56);/q;+1/p-1/b13-11-,19-17-,24-22-,30-28-;/t39-,42?,43-,44?,45?,46?,47-;/m1./s1

InChI key

NHSOICFDOXKHEC-ZGISKSNKSA-M

General description

Phosphatidylinositol (PI) is a phosphoglyceride, which has an inositol head group. Liver PI is a natural phosphatidylinositol. PI is the most important phospholipid component of the plasma membrane.

Application

Liver PI has been used as a standard for the identification of chromatogram of phospholipid (PL) classes in soy extract. It has also been used for the preparation of liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (840042P-100mg)
5 mL Clear Glass Sealed Ampule (840042P-10mg)
5 mL Clear Glass Sealed Ampule (840042P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phosphatidylethanolamine molecular species in various food matrices by liquid chromatography--electrospray ionization--tandem mass spectrometry (LC--ESI--MS 2)
Zhou L, et al.
Analytical and Bioanalytical Chemistry, 403(1) (2012)
Molecular Mechanisms of Resistance to Agrochemicals (2000)
Cell Biology (2007)
Ruzica Bago et al.
The Biochemical journal, 463(3), 413-427 (2014-09-02)
The Vps34 (vacuolar protein sorting 34) class III PI3K (phosphoinositide 3-kinase) phosphorylates PtdIns (phosphatidylinositol) at endosomal membranes to generate PtdIns(3)P that regulates membrane trafficking processes via its ability to recruit a subset of proteins possessing PtdIns(3)P-binding PX (phox homology) and
Examination of the Interaction between a Membrane Active Peptide and Artificial Bilayers by Dual Polarisation Interferometry
Payne JAE, et al.
Bio-protocol, 7(1), e2087-e2087 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service