Skip to Content
Merck
All Photos(3)

Documents

109959

Sigma-Aldrich

2-Ethylbutyric acid

99%

Synonym(s):

Diethylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2CHCO2H
CAS Number:
Molecular Weight:
116.16
Beilstein:
1098634
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.08 mmHg ( 20 °C)

Assay

99%

refractive index

n20/D 1.413 (lit.)

bp

99-101 °C/18 mmHg (lit.)

mp

−16-−13 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

CCC(CC)C(O)=O

InChI

1S/C6H12O2/c1-3-5(4-2)6(7)8/h5H,3-4H2,1-2H3,(H,7,8)

InChI key

OXQGTIUCKGYOAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethylbutyric acid is used as the internal standard for determining volatile fatty acids (VFAs) in wastewater from the constructed wetlands. It is used in the determination of both free and fatty acids in milk and milk products by HPLC method with direct derivatisation.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Blood,Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Liu et al.
Food & function, 7(9), 3772-3781 (2016-10-08)
Indigestible resistant starches (RS) are substrates for gut-microbial metabolism and have been shown to attenuate intestinal inflammation but the supporting evidence is inconsistent and lacks mechanistic explanation. We have recently reported dietary RS type 4 (RS4) induced improvements in immunometabolic
Lin Feng et al.
Journal of chromatographic science, 46(7), 577-584 (2008-08-23)
A headspace solid-phase microextraction (HS-SPME) followed by gas chromatography coupled to flame ionization detector (GC-FID) is developed for the direct determination of volatile fatty acids (VFAs) in wastewater from constructed wetlands. Through the investigation on extraction behavior of different coatings
Evdokia K Mitsou et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-24)
Alterations of gut microbiota are evident during the aging process. Prebiotics may restore the gut microbial balance, with β-glucans emerging as prebiotic candidates. This study aimed to investigate the impact of edible mushrooms rich in β-glucans on the gut microbiota
Evangelia N Kerezoudi et al.
Food & function, 12(4), 1529-1546 (2021-02-02)
Recent data have highlighted the role of the gut microbiota and its several metabolites in maintaining bone health. Thus, gut microbiota manipulation, e.g., by prebiotics, might offer a plausible target in the fight against bone degenerative diseases. This study aimed
Faiga Magzal et al.
Scientific reports, 11(1), 4052-4052 (2021-02-20)
Insomnia is a disorder characterized by difficulty falling asleep and poor sleep continuity and is associated with increased risks for physical and cognitive decline. Insomnia with short sleep duration is considered the most biologically severe phenotype of the disorder. Evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service