Skip to Content
Merck
All Photos(1)

Key Documents

W278430

Sigma-Aldrich

Nonanoic acid

natural, 98%, FG

Synonym(s):

Acid C9, Pelargonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COOH
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2784
Beilstein:
1752351
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.029
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
Olea europaea
food allergen:
no known allergens

biological source

Olea europaea

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 173.315
FDA 21 CFR 178.1010

vapor density

5.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

98%

expl. lim.

9 %

refractive index

n20/D 1.432 (lit.)

bp

268-269 °C (lit.)

mp

9 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cheesy; waxy; tallow

SMILES string

CCCCCCCCC(O)=O

InChI

1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)

InChI key

FBUKVWPVBMHYJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilbur Johnson et al.
International journal of toxicology, 30(6 Suppl), 228S-269S (2012-01-25)
Pelargonic acid and its esters function as skin-conditioning agents in cosmetics. Molecular weight (mw) and octanol-water partition coefficient data suggest that dermal penetration is possible. The biohandling of branched-chain fatty acids is not the same as for straight-chain fatty acids
Jan E Wahlberg et al.
Contact dermatitis, 49(3), 117-123 (2003-12-18)
Irritant contact dermatitis is defined as a non-immunological skin reaction following exposure to various chemical, mechanical and physical factors. It is known that the skin response to irritants depends on the irritant applied and differs between chemically different irritants. Sodium
J Wang et al.
Journal of dairy science, 104(2), 1576-1590 (2020-12-29)
To determine the odor-active compounds in Cheddar cheeses with different ripening times (6, 10, and 14 mo), 39 potent odorants of Cheddar cheeses were identified with a flavor dilution factor range between 1 and 512 by aroma extract dilution analysis.
Lulu Wang et al.
Journal of agricultural and food chemistry, 68(6), 1666-1677 (2020-01-21)
A potent unpleasant aroma presenting a pickle-like off-odor in Moutai-aroma type Baijiu was studied by comparative aroma extract dilution analysis (AEDA). Two Moutai-aroma type Baijiu samples, one having no off-odor (A) and the other one presenting pickle-like off-odor (B), were
Raluca Ciuraru et al.
Scientific reports, 5, 12741-12741 (2015-08-06)
The sea-surface microlayer (SML) has different physical, chemical and biological properties compared to the subsurface water, with an enrichment of organic matter i.e., dissolved organic matter including UV absorbing humic substances, fatty acids and many others. Here we present experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service