Skip to Content
Merck
All Photos(1)

Key Documents

W278408

Sigma-Aldrich

Nonanoic acid

≥96%, FG

Synonym(s):

Acid C9, Pelargonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COOH
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2784
Beilstein:
1752351
EC Number:
Council of Europe no.:
29
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.029
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 1010
FDA 21 CFR 172.515
FDA 21 CFR 315

vapor density

5.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥96%

expl. lim.

9 %

refractive index

n20/D 1.432 (lit.)

bp

268-269 °C (lit.)

mp

9 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

waxy; cheesy; tallow

SMILES string

CCCCCCCCC(O)=O

InChI

1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)

InChI key

FBUKVWPVBMHYJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonanoic acid occurs naturally in Pelargonium L′Her.[1] It one of the flavor constituents of cooked rice[2], acerola fruit[3], licorice[4] and yogurt.[5]

Application

Nonanoic acid has been used as a standard for the quantification of odorant compounds in French mimosa absolute oil by GC-Sniffing.[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile flavor components of licorice.
Frattini C, et al.
Journal of Agricultural and Food Chemistry, 25(6), 1238-1241 (1977)
Alternative management of roadside vegetation.
Barker AV & Prostak RG.
HortTechnology, 19(2), 346-352 (2009)
Chemical composition of French mimosa absolute oil.
Perriot R, et al.
Journal of Agricultural and Food Chemistry, 58(3), 1844-1849 (2010)
Volatile flavor compounds in yogurt: a review
Cheng H.
Critical Reviews in Food Science and Nutrition, 50(10), 938-950 (2010)
Zhiyong Sun et al.
Applied microbiology and biotechnology, 74(1), 69-77 (2006-10-26)
Pseudomonas putida KT2440 grew on glucose at a specific rate of 0.48 h(-1) but accumulated almost no poly-3-hydroxyalkanoates (PHA). Subsequent nitrogen limitation on nonanoic acid resulted in the accumulation of only 27% medium-chain-length PHA (MCL-PHA). In contrast, exponential nonanoic acid-limited

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service