Skip to Content
Merck
All Photos(1)

Key Documents

W268313

Sigma-Aldrich

Methyl benzoate

natural, ≥98%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15
FEMA Number:
2683
Beilstein:
1072099
EC Number:
Council of Europe no.:
260c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.725
NACRES:
NA.21
Organoleptic:
floral; phenolic
grade:
FG
Fragrance grade
Halal
Kosher
natural
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

4.68 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.516 (lit.)

bp

198-199 °C (lit.)

mp

−12 °C (lit.)

solubility

ethanol: 60%, clear (1mL/4ml)

density

1.088 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

floral; phenolic

SMILES string

COC(=O)c1ccccc1

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

QPJVMBTYPHYUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl benzoate occurs naturally in guava, mango and kiwifruit.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of headspace solid-phase microextraction to volatile flavour profile development during storage and ripening of kiwifruit.
Wan XM, et al.
Food Research International, 32(3), 175-183 (1999)
Dynamic headspace concentration and gas chromatography of volatile flavor components in peach.
Narain N, et al.
Journal of Food Science, 55(5), 1303-1307 (1990)
Contribution of volatile compounds to mango (Mangifera indica L.) aroma
Pino JA & Mesa J
Flavour and Fragrance Journal, 21(2), 207-213 (2006)
María Manchado-Rojo et al.
The Plant journal : for cell and molecular biology, 72(2), 294-307 (2012-06-20)
The transcriptional network topology of B function in Antirrhinum, required for petal and stamen development, is thought to rely on initial activation of transcription of DEFICIENS (DEF) and GLOBOSA (GLO), followed by a positive autoregulatory loop maintaining gene expression levels.
Nan Zhao et al.
Plant physiology and biochemistry : PPB, 48(4), 279-287 (2010-03-05)
Two benzenoid esters, methyl salicylate (MeSA) and methyl benzoate (MeBA), were detected from insect-damaged rice plants. By correlating metabolite production with gene expression analysis, five candidate genes encoding putative carboxyl methyltransferases were identified. Enzymatic assays with Escherichia coli-expressed recombinant proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service