Skip to Content
Merck
All Photos(1)

Key Documents

B6630

Sigma-Aldrich

Benzyl benzoate

ReagentPlus®, ≥99.0%

Synonym(s):

Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
2049280
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1 mmHg ( 125 °C)

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

liquid

autoignition temp.

896 °F

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

density

1.118 g/mL at 20 °C (lit.)

functional group

ester
phenyl

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzyl benzoate is a benzyl compound that can be synthesized by reacting benzyl chloride with sodium benzoate in the presence of tetrabutylaramonium iodide. It is reported to be the key constituent in the essential oils isolated from leaves and stem bark of Cinnamomum zeylanicum. Its toxicity has been assessed. The mechanism of thermal decomposition of benzyl benzoate has been investigated.

Application

Benzyl benzoate has been used to:
  • Prepare benzyl alcohol/benzyl benzoate (BABB) solution employed for enhancing the tissue transparency during the whole-mount immunostaining BABB method.
  • Study its ability to induce an olfactory response in the third instar larvae of Drosophila melanogaster.
  • Prepare Spalteholz fluid in combination with methyl salicylate, employed during LSFM (light sheet fluorescence microscopy) imaging.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jos van Rooij et al.
Biomedical optics express, 10(4), 1782-1793 (2019-05-16)
In this work we demonstrate large-scale high-sensitivity optical diffraction tomography (ODT) of zebrafish. We make this possible by three improvements. First, we obtain a large field of view while still maintaining a high resolution by using a high magnification over
Pyrolytic Chemistry of Benzyl Benzoate.
Chen PS and Chou CH.
J. Chin. Chem. Soc., 41(5), 597-599 (1994)
Toxicity studies on benzyl benzoate and related benzyl compounds.
B E GRAHAM et al.
The Journal of pharmacology and experimental therapeutics, 84, 358-362 (1945-08-01)
Kinetics of phase transfer catalytic preparation of benzyl benzoate.
Chang JR, et al.
J. Chin. Chem. Soc., 31(2), 185-190 (1984)
Volume shrinkage of bone, brain and muscle tissue in sample preparation for micro-CT and light sheet fluorescence microscopy (LSFM).
Buytaert J, et al.
Microscopy and Microanalysis, 20(04), 1208-1217 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service