Skip to Content
Merck
All Photos(1)

Documents

T24201

Sigma-Aldrich

Tetramethylthiuram disulfide

97%

Synonym(s):

Bis(dimethylthiocarbamoyl) disulfide, Bis(dimethylthiocarbamyl) disulfide, Thiram

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCSS2CSN(CH3)2
CAS Number:
Molecular Weight:
240.43
Beilstein:
1725821
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

autoignition temp.

316 °F

mp

156-158 °C (lit.)

SMILES string

CN(C)C(=S)SSC(=S)N(C)C

InChI

1S/C6H12N2S4/c1-7(2)5(9)11-12-6(10)8(3)4/h1-4H3

InChI key

KUAZQDVKQLNFPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiwei Deng et al.
PloS one, 10(6), e0129781-e0129781 (2015-06-13)
The plant circadian clock is an internal timekeeper that coordinates biological processes with daily changes in the external environment. The transcript levels of clock genes, which oscillate to control circadian outputs, were examined during early seedling development in barley (Hordeum
Agnese D'Agostino et al.
Talanta, 216, 120936-120936 (2020-05-28)
Seed-growth synthesis is a common strategy to prepare silver nanoplates, whose peculiar plasmonic features can be exploited for surface enhanced Raman scattering (SERS) applications. Here we describe the fabrication and characterization of SERS chips using a peculiar in situ seed
Vaneet Kumar Sharma et al.
Journal of environmental monitoring : JEM, 5(5), 717-723 (2003-11-01)
In this review a brief introduction to thiram (tetramethylthiuram disulfide; TMTD) pesticide has been given along with other applications. All the important methods available are systematically arranged and are listed under various techniques. Some of these methods have been applied
Toxicology of thiram (tetramethylthiuram disulfide): a review.
R R Dalvi
Veterinary and human toxicology, 30(5), 480-482 (1988-10-01)
Olga M S Filipe et al.
Chemosphere, 90(2), 432-440 (2012-09-07)
This work aimed to assess the influence of soil copper content on the sorption processes of thiram, a fungicide widely used in agriculture, most of the times together with copper. Two different types of studies were performed: (1) desorption studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service