Skip to Content
Merck
All Photos(2)

Key Documents

D207756

Sigma-Aldrich

1,3-Diphenylguanidine

97%

Synonym(s):

1,2-Diphenylguanidine, N,N′-Diphenylguanidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5NH)2C=NH
CAS Number:
Molecular Weight:
211.26
Beilstein:
1875653
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

146-148 °C (lit.)

SMILES string

N=C(Nc1ccccc1)Nc2ccccc2

InChI

1S/C13H13N3/c14-13(15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H,(H3,14,15,16)

InChI key

OWRCNXZUPFZXOS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occupational allergic contact dermatitis from diphenylguanidine in a gas mask.
M Bruze et al.
Contact dermatitis, 31(2), 125-126 (1994-08-01)
Mingxiang Lin et al.
Journal of pharmaceutical and biomedical analysis, 45(5), 747-755 (2007-10-12)
During the content uniformity test of a drug product (tablet formulation), an unknown peak was observed in the HPLC chromatograms. Upon further investigation, it was determined that the unknown peak was originated from an external source and, therefore, the drug
P V Shah et al.
Journal of toxicology and environmental health, 15(5), 623-633 (1985-01-01)
Dermal absorption, distribution, and metabolism of 1,3-diphenylguanidine (CAS 102-06-7) (DPG), widely used as an accelerator in processing rubber and in food packaging, was studied in adult female Sprague-Dawley rats. DPG shows 10% penetration through clipped back skin of the rats
Allergy to rubber additives in orthopedic braces.
S T Bolhaar et al.
Contact dermatitis, 42(5), 288-289 (2000-05-02)
Lymphomatoid allergic contact dermatitis mimicking cutaneous T cell lymphoma.
Meghan T Hession et al.
Dermatitis : contact, atopic, occupational, drug, 21(4), 220-220 (2010-07-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service