Skip to Content
Merck
All Photos(2)

Key Documents

567205

Sigma-Aldrich

Tetramethylthiuram monosulfide

97%

Synonym(s):

UNADS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((H3C)2NCS)2S
CAS Number:
Molecular Weight:
208.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

107-111 °C (lit.)

functional group

amine
dithiocarbamate
thioether

SMILES string

CN(C)C(=S)SC(=S)N(C)C

InChI

1S/C6H12N2S3/c1-7(2)5(9)11-6(10)8(3)4/h1-4H3

InChI key

REQPQFUJGGOFQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

312.8 °F - closed cup

Flash Point(C)

156 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Contact allergy to Nobecutan.
B Pock-Steen
Contact dermatitis, 18(1), 52-53 (1988-01-01)
A Rannug et al.
Chemico-biological interactions, 49(3), 329-340 (1984-05-01)
In recent years data have accumulated regarding genotoxic properties of dithiocarbamic acid derivatives. The results from the present work indicate that the mutagenicity of these compounds depends on an indirect effect via oxygen radicals. Mutagenicity of tetramethylthiuram disulfide ( TMTD
B B Knudsen et al.
Contact dermatitis, 34(6), 410-413 (1996-06-01)
An estimate of amounts of thiurams that may be released from rubber gloves into synthetic sweat, has previously been generated. These amounts should be compared to elicitation thresholds of patch tests performed with serial dilutions of thiuram mix using synthetic
Q Li et al.
Toxicology, 104(1-3), 17-23 (1995-12-15)
To examine the validity of a modified lymphocyte transformation test for evaluating contact hypersensitivity from weak sensitizers, guinea pigs were sensitized with formaldehyde (F) or tetramethylthiuram monosulfide (TMTM) using the maximization test procedure. Lymph node cells from the animals were
G García de Torres et al.
Drug and chemical toxicology, 6(4), 317-328 (1983-01-01)
In adult female Wistar rats, pretreated by gavage with two doses - 16 or 256 mumol/kg - of cyanamide, TMTD (tetramethylthiuram disulfide), TMTM (tetramethylthiuram monosulfide), Ziram or Zineb at 90 min or 18 h before administration of 2 g of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service