Skip to Content
Merck
All Photos(2)

Key Documents

329703

Sigma-Aldrich

Zinc diethyldithiocarbamate

97%

Synonym(s):

Zinc bis(diethyldithiocarbamate), Zinc diethylcarbamodithioate, Diethyldithiocarbamic acid zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C2H5)2NCS2]2Zn
CAS Number:
Molecular Weight:
361.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

core: zinc
reagent type: catalyst

mp

178-181 °C (lit.)

SMILES string

CCN(CC)C(=S)S[Zn]SC(=S)N(CC)CC

InChI

1S/2C5H11NS2.Zn/c2*1-3-6(4-2)5(7)8;/h2*3-4H2,1-2H3,(H,7,8);/q;;+2/p-2

InChI key

RKQOSDAEEGPRER-UHFFFAOYSA-L

General description

Zinc diethyldithiocarbamate is an organometallic precursor used as an accelerator in the vulcanization of natural rubber latex films. [1] [2]

Application

Zinc diethyldithiocarbamate (Zn(S2CNEt2)2, ZDTC) can be used as an organocatalyst for the synthesis of high molecular weight polyurethanes from PEG- and PPG-diol macromonomers for use in biomedical applications. In comparison to other zinc catalysts, ZDTC demonstrated outstanding activity in the preparation of polyurethanes because of its tolerance to different organic solvents.[3]
It can also be used as:
  • A catalyst for the polymerization of olefin oxides.[4]
  • A precursor to prepare zinc sulfide semiconductor nanowires, which can be used as building blocks for photonic devices.[5]

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Liver,spleen, Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zinc diethyldithiocarbamate as a catalyst for synthesising biomedically-relevant thermogelling polyurethanes
Lim JYC, et al.
Materials Advances, 1(9), 3221-3232 (2020)
Epitaxial ZnS thin films grown by single source chemical vapor deposition
Nguyen T H et al.
The Journal of Physical Chemistry B, 104, 1150-1152 (2000)
Characterization of the natural rubber vulcanizates obtained by different accelerators
J Travas-Sejdic et al.
European Polymer Journal, 32, 1395-1401 (1996)
Polymerization of olefin oxides with zinc n-butyl xanthate and zinc dimethyldithiocarbamate catalysts: Mechanism of initiation
L Joginder and Devlin EF
Journal of Polymer Science, 11(1), 119-133 (1973)
Synthesis of CdS and ZnS nanowires using single-source molecular precursors
Barrelet CJ, et al.
Journal of the American Chemical Society, 125(38), 11498-11499 (2003)

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is this product steril? if not, what is the optimum sterilization method?

    1 answer
    1. The product is not sterile. Unfortunately, a sterilization technique is not determined.

      Helpful?

  4. Is this product available as sterile films CAS136-23-2 and CAS 14324-55-1

    1 answer
    1. These products are currently not offered as sterile films.

      Helpful?

  5. How can I get the expiry date for product: 329703-25G bought from Sigma?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service