Skip to Content
Merck
All Photos(1)

Key Documents

T111

Sigma-Aldrich

8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride

≥98%

Synonym(s):

TMB 8 hydrochloride, TMB-8 · HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CO2(CH2)8N(C2H5)2 · HCl
CAS Number:
Molecular Weight:
431.99
Beilstein:
5466611
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

94-97 °C (lit.)

SMILES string

Cl[H].CCN(CC)CCCCCCCCOC(=O)c1cc(OC)c(OC)c(OC)c1

InChI

1S/C22H37NO5.ClH/c1-6-23(7-2)14-12-10-8-9-11-13-15-28-22(24)18-16-19(25-3)21(27-5)20(17-18)26-4;/h16-17H,6-15H2,1-5H3;1H

InChI key

KFJZVXKPPQIYCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Pharmacologically active inhibitor of hepatitis C virus life cycle

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrycja Libako et al.
Magnesium research, 25(1), 12-20 (2012-03-30)
Divalent cations, especially calcium and magnesium, have been shown to play an important regulatory role in endothelial and immune cells. To learn more about the interaction of these two metals in the regulation of cell growth, we altered the calcium/magnesium
Nancy S Krieger et al.
American journal of physiology. Renal physiology, 300(1), F91-F97 (2010-11-05)
In vivo chronic metabolic acidosis induces net Ca2+ efflux from bone, and incubation of neonatal mouse calvariae in medium simulating physiological metabolic acidosis induces bone resorption. It appears that activation of the proton (H+) receptor OGR1 in the osteoblast leads
Hai-Long Wang et al.
The Chinese journal of physiology, 55(2), 101-107 (2012-05-09)
The purpose of this study was to determine the effects of an extract from Moringa oleifera (MO) on the development of monocrotaline (MCT)-induced pulmonary hypertension (PH) in Wistar rats. An ethanol extraction was performed on dried MO leaves, and HPLC
María Angeles Sánchez-Sampedro et al.
Journal of plant physiology, 162(10), 1177-1182 (2005-11-01)
Elimination of calcium ions from the medium of cell cultures of Silybum marianum (L.) Gaertn increased flavonolignan production. Silymarin accumulation was not altered by treatment of cultures with the calcium ionophore A23187. The specific Ca2+ chelator, EGTA, enhanced the silymarin
Jafar Vatanparast et al.
European journal of pharmacology, 571(2-3), 81-87 (2007-06-30)
We have previously reported that paraoxon, an organophosphate compound, at submicromolar concentrations effectively suppresses Ca2+ action potentials and modulates the activity of snail neurons. This effect was unrelated to acetylcholinesterase inhibition but was found to involve the direct or indirect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service