Skip to Content
Merck
All Photos(2)

Documents

O9328

Sigma-Aldrich

Oxamide

98%

Synonym(s):

Oxalic acid diamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COCONH2
CAS Number:
Molecular Weight:
88.07
Beilstein:
1743262
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

>300 °C (lit.)

SMILES string

NC(=O)C(N)=O

InChI

1S/C2H4N2O2/c3-1(5)2(4)6/h(H2,3,5)(H2,4,6)

InChI key

YIKSCQDJHCMVMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxamide can be used:
  • As a precursor for the synthesis of ligands such as bis(benzimidazole) and Schiff base ligands formed by condensation with furfural.
  • Carbon nitride (g-C3N4) nanotubes by self-assembly polymerization with urea.
  • As a bridging ligand for the synthesis of binuclear IrIII complex [Ir22-oxamidato-N,N′,O,O′)(ptpy)4], ptpy = 2-(p-tolyl)pyridinato.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Na Zhu et al.
Inorganic chemistry, 46(4), 1297-1304 (2007-01-25)
Four heteronuclear complexes Mn(CuL)2(SCN)2 (1), {[Mn(CuL)2(mu-dca)2].2H2O}n (2), Zn(CuL)2(SCN)2 (3), and [Fe(CuL)(N3)2]2 (4) incorporating macrocyclic oxamide ligands have been synthesized and structurally characterized. L is the dianion of diethyl 5,6,7,8,15,16-hexahydro-6,7-dioxodibenzo[1,4,8,11]-tetraazacyclotetradecine-13,18-dicarboxylate, and dca is the dicyanamide. The structure of 1 or 3
Sergey P Gavrish et al.
Dalton transactions (Cambridge, England : 2003), (41)(41), 4708-4714 (2007-10-18)
A comparison of the molecular structure of related nickel(II) complexes of the open-chain and 13-membered macrocyclic oxamide-derived ligands NiL(1).4H2O and NiL(2).3H2O revealed that the formation of an additional 6-membered chelate ring in the complex results in rather small changes in
Synthesis and Molecular Structure of the New Green Emitting Complex [Ir2(?2-oxamidato-N, N?, O, O?)(2-(p-tolyl)pyridinato)4]
Graf M, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 639(7), 1090-1094 (2013)
Yun Fei Long et al.
Analytica chimica acta, 624(1), 128-132 (2008-08-19)
Light scattering (LS) signals have been applied for analytical detections, but the selectivity is poor. In order to improve the selectivity, pre-separation or new machines are generally considered. Differing from these methods, we synthesized a highly selective oxamide ligand, N',N'-bis
Bis (benzimidazole) as supramolecular building block in manganese (IV) chemistry
Samol'ova E, et al.
Journal of Molecular Structure, 1176, 366-375 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service