Skip to Content
Merck
All Photos(2)

Key Documents

292893

Sigma-Aldrich

Adipamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCO(CH2)4CONH2
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

226-229 °C (lit.)

solubility

formic acid: soluble 50 mg/mL, clear, colorless to faintly yellow

functional group

amide

SMILES string

NC(=O)CCCCC(N)=O

InChI

1S/C6H12N2O2/c7-5(9)3-1-2-4-6(8)10/h1-4H2,(H2,7,9)(H2,8,10)

InChI key

GVNWZKBFMFUVNX-UHFFFAOYSA-N

Application

Adipamide has been used in the preparation of bisimidates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Morris et al.
The Biochemical journal, 147(3), 593-603 (1975-06-01)
Triethyloxonium tetrafluoroborate was used to O-alkylate nylon-tube thus producing the imidate salt of the nylon which was further made to react with 1,6-diaminohexane. 2. Hexokinase (EC 2.7.1.1) and glucose 6-phosphate dehydrogenase (EC 1.1.1.49) were immobilized on the amino-substituted nylon tube
S J Kennel et al.
Toxicology and applied pharmacology, 82(2), 256-263 (1986-02-01)
A thyroglobulin conjugate of dioxin (thyroglobulin-2 adipamide, 3,7,8-trichlorodibenzo-p-dioxin) (TG-TCDD) was used to immunize BALB/c mice. Hybridomas were produced by cell fusion between immune spleen cells and mouse myelomas SP2/0, P3, or NS1. To screen the thousands of resultant cultures for
R W Fleischman et al.
Journal of environmental pathology and toxicology, 3(5-6), 149-170 (1980-06-01)
As a part of the National Cancer Institute's effort to screen environmental and occupational chemicals for chronic toxicity and carcinogenicity, the amides acetamide, hexanamide, adipamide, urea, and p-tolylurea were fed to male and female C57B1/6 mice and Fischer 344 rats
Xuejiang Wang et al.
Biomaterials, 23(24), 4787-4791 (2002-10-04)
In this study, acicular nano-hydroxyapatite (n-HA) was used to make a new biomimetic composite with polyamide (poly hexamethylene adipamide) by a unique technique. The physical and chemical characteristics of the composites were tested. It was found that these synthesized n-HA
Colin C Seaton et al.
Chemical communications (Cambridge, England), (8)(8), 880-881 (2002-07-19)
The crystal structure of a previously unknown triclinic polymorph of adipamide has been solved from laboratory X-ray powder diffraction data using a new direct space global optimisation method based on differential evolution.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service