Skip to Content
Merck
All Photos(3)

Documents

195464

Sigma-Aldrich

2,5-Dihydroxy-1,4-benzoquinone

98%

Synonym(s):

2,5-Dihydroxy-2,5-cyclohexadiene-1,4-dione, 2,5-Dihydroxy-p -benzoquinone, 2,5-Dihydroxycyclohexa-2,5-diene-1,4-dione, Anilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H2(=O)2
CAS Number:
Molecular Weight:
140.09
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

235 °C (dec.) (lit.)

SMILES string

OC1=CC(=O)C(O)=CC1=O

InChI

1S/C6H4O4/c7-3-1-4(8)6(10)2-5(3)9/h1-2,7,10H

InChI key

QFSYADJLNBHAKO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengxia Chen et al.
Chemosphere, 250, 126226-126226 (2020-02-24)
A novel catalyst which integrates heterogeneous and homogenous Fenton reactions is designed and fabricated by encapsulating 2,5-dihydroxy-1,4-benzoquinone (2,5-DBQ) in ECDP-Fe3O4, a composite of Fe3O4 nanoparticles immobilized on a β-cyclodextrin polymer (ECDP) with ethylene diamine tetraacetic acid (EDTA) as cross-linking agent.
Nathalie Honorio-Felício et al.
Journal of inorganic biochemistry, 164, 34-41 (2016-09-12)
Conformational changes associated to sensing mechanisms of heme-based protein sensors are a key molecular event that seems to modulate not only the protein activity but also the potential of the Fe
M Ahmed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56(5), 965-981 (2000-05-16)
The electronic absorption spectra of 1,4-benzoquinone (BQ) and its 2,5-dihydroxy and tetrahydroxy derivatives have been studied in detail. The interpretation of the electronic bands is made on the basis of PPP and CNDO calculations. It is found that the pi
Jan Bohuslavek et al.
Biodegradation, 16(4), 353-362 (2005-05-04)
Hydroxyquinol, a common metabolite of aromatic compounds, is readily auto-oxidized to hydroxyquinone. Enzymatic activities that metabolized hydroxyquinone were observed from the cell extracts of Sphingobium chlorophenolicum ATCC 39723. An enzyme capable of transforming hydroxyquinone was partially purified, and its activities
Y Sun et al.
Guang pu xue yu guang pu fen xi = Guang pu, 20(6), 790-792 (2003-08-27)
In this paper, we report the Ab initio calculation and experimental investigation of the vibrational frequencies of 4,5-dihydroxy-1,2-benzoquinone and 2,5-dihydroxy-1,4-benzoquinone. The main IR and Raman absorption bands of them have been assigned, learned from other strong points to offset one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service