All Photos(1)
About This Item
Linear Formula:
C5H7CH3
CAS Number:
Molecular Weight:
82.14
Beilstein:
1900640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
bp
72 °C/754 mmHg (lit.)
density
0.78 g/mL at 25 °C (lit.)
SMILES string
CC1=CCCC1
InChI
1S/C6H10/c1-6-4-2-3-5-6/h4H,2-3,5H2,1H3
InChI key
ATQUFXWBVZUTKO-UHFFFAOYSA-N
Related Categories
Application
1-Methylcyclopentene can be used as:
- A probe molecule in the synthesis of alkenyl carbenium ions via adsorption on zeolite Y.
- A model olefin compound in the kinetic study of hydrogenation of olefins using various catalytic systems.
- A reactant to synthesize 1,2,3-trisubstituted benzocyclopentenes by reacting with β-disulfonyl iodonium ylides.
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-2.2 °F - closed cup
Flash Point(C)
-19 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Formation of stable alkenyl carbenium ions in high yield by adsorption of 1-methylcyclopentene on zeolite Y at low temperature
Yang S, et al.
Chemical Communications (Cambridge, England), 2008-2009 (2001)
A facile diastereoselective synthesis of functionalized 1, 2, 3-trisubstituted benzocyclopentenes through the cycloaddition of bis (phenylsulfonyl) iodonium ylides to cyclic alkenes
Adam W, et al.
The Journal of Organic Chemistry, 68, 9155-9158 (2003)
Kinetic study of olefin hydrogenation on hydrotreating catalysts
Badawi M, et al.
J. Mol. Catal. A: Chem., 320, 34-39 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service