Skip to Content
Merck
All Photos(1)

Key Documents

M67303

Sigma-Aldrich

2-Methyl-2-pentene

98%

Synonym(s):

2M2P

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1731107
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

267 mmHg ( 100 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.400 (lit.)

bp

67 °C (lit.)

mp

−135 °C (lit.)

density

0.69 g/mL at 25 °C (lit.)

SMILES string

CC\C=C(\C)C

InChI

1S/C6H12/c1-4-5-6(2)3/h5H,4H2,1-3H3

InChI key

JMMZCWZIJXAGKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-2-pentene has been used in various photochemical and ozonolysis studies.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Production of stabilized Criegee intermediates and peroxides in the gas phase ozonolysis of alkenes: 1. Ethene, trans?2?butene, and 2, 3?dimethyl?2?butene.
Hasson AS, et al.
Journal of Geophysical Research: Atmospheres, 106(D24), 34131-34142 (2001)
The gas-phase ozonolysis of unsaturated volatile organic compounds in the troposphere.
Johnson D and Marston G
Chemical Society Reviews, 37(4), 699-716 (2008)
Production and quenching of singlet oxygen by the sensitizer in dye-sensitized photo-oxygenations.
Tanielian C, et al.
Journal of Photochemistry, 25(2-4), 117-125 (1984)
Chemistry of singlet oxygen. 44. Mechanism of photooxidation of 2, 5-dimethylhexa-2, 4-diene and 2-methyl-2-pentene.
Manring LE and Foote CS
Journal of the American Chemical Society, 105(14), 4710-4717 (1983)
Gas?phase ozonolysis: Rate coefficients for a series of terpenes and rate coefficients and OH yields for 2?methyl?2?butene and 2, 3?dimethyl?2?butene.
Witter M, et al.
International Journal of Chemical Kinetics, 34(6), 394-403 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service