Skip to Content
Merck
All Photos(3)

Key Documents

M39407

Sigma-Aldrich

Methylcyclopentane

97%

Synonym(s):

Methycyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9CH3
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900214
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

232.8 mmHg ( 37.7 °C)

Assay

97%

autoignition temp.

624 °F

expl. lim.

8.35 %

refractive index

n20/D 1.409 (lit.)

bp

72 °C (lit.)

mp

−142 °C (lit.)

density

0.749 g/mL at 25 °C (lit.)

SMILES string

CC1CCCC1

InChI

1S/C6H12/c1-6-4-2-3-5-6/h6H,2-5H2,1H3

InChI key

GDOPTJXRTPNYNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methylcyclopentane can undergo ring opening or ring enlargement to yield various hydrocarbons including branched and unbranched hexanes, cyclohexane and benzene.[1][2][3]

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of acidic properties of mesoporous zeolites supporting pt nanoparticles on hydrogenative conversion of methylcyclopentane.
Na, Kyungsu et al.
Journal of the American Chemical Society, 136(49), 17207-17212 (2014)
Size and shape dependence on Pt nanoparticles for the methylcyclopentane/hydrogen ring opening/ring enlargement reaction.
Alayoglu, S et al.
Catalysis Letters, 141(7), 914-924 (2011)
Ring-opening reactions of methylcyclopentane over metal catalysts, M= Pt, Rh, Ir, and Pd: A mechanistic study from first-principles calculations.
Zhao, Zhi-Jian et al.
ACS Catalysis, 3(2), 196-205 (2013)
Haijing Wang et al.
Journal of chromatographic science, 53(5), 647-654 (2014-09-17)
Determination of volatile organic compounds (VOCs) in crude oil, such as super volatile organic compounds (super VOCs) and simple polycyclic aromatic hydrocarbons (PAHs), is vital for targeting crude oil spill spots. In this study, a static headspace gas chromatography flame
Yulia Kaluzhny et al.
Alternatives to laboratory animals : ATLA, 43(2), 101-127 (2015-05-23)
The 7th Amendment to the EU Cosmetics Directive and the EU REACH Regulation have reinforced the need for in vitro ocular test methods. Validated in vitro ocular toxicity tests that can predict the human response to chemicals, cosmetics and other

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service