Skip to Content
Merck
All Photos(1)

Documents

111015

Sigma-Aldrich

Methylenecyclohexane

98%

Synonym(s):

1-Methylenecyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=CH2)
CAS Number:
Molecular Weight:
96.17
Beilstein:
773760
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

102-103 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=C1CCCCC1

InChI

1S/C7H12/c1-7-5-3-2-4-6-7/h1-6H2

InChI key

YULMNMJFAZWLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylenecyclohexane ring is obtained by 1,4-addition of 4-pentenylmagnesium bromide to 2-cyano-2-cycloalkenones followed by a Pd(II)-mediated oxidative cyclization. It undergoes gas phase ozonolysis to form secondary organic aerosol (SOA).

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conny Müller et al.
Electrophoresis, 28(9), 1364-1370 (2007-03-21)
The composition of secondary organic aerosol (SOA) from the gas phase ozonolysis of methylenecyclohexane was analyzed in a series of indoor aerosol chamber experiments. Capillary electrophoresis-electrospray ionization-ion trap mass spectrometry (CE/ESI-ITMS) was used for qualitative and quantitative analysis of SOA
Min-Tsang Hsieh et al.
Organic & biomolecular chemistry, 10(23), 4609-4617 (2012-05-15)
A highly efficient annulative approach towards the construction of the structurally attractive methylenecyclohexane ring was developed through a convenient 1,4-addition of 4-pentenylmagnesium bromide to 2-cyano-2-cycloalkenones followed by a Pd(II)-mediated oxidative cyclization of the resulting ω-unsaturated α-cyano ketones. Based on this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service